Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 107378362-107378603 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC54
Entrez Gene ID
84692
Gene Name
coiled-coil domain containing 54
Gene Aliases
NYD-SP17, SP17
UniGene
Hs.164799
Gene Chromosome Location
Chr.3: 107377341-107378634 on Build GRCh38
Transcripts
NM_032600.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6095846 c.792T>G p.S264R Chr.3: 107378379-107378379 on Build GRCh38
4389451 c.914C>T p.T305I Chr.3: 107378501-107378501 on Build GRCh38
2916520 c.946G>A p.G316S Chr.3: 107378533-107378533 on Build GRCh38
727456 c.798C>A p.T266T Chr.3: 107378385-107378385 on Build GRCh38
4112023 c.972A>C p.E324D Chr.3: 107378559-107378559 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs557598246 Chr.3: 107378443-107378443 on Build GRCh38
rs577435697 Chr.3: 107378480-107378480 on Build GRCh38
rs765197465 Chr.3: 107378413-107378413 on Build GRCh38
rs912128893 Chr.3: 107378407-107378407 on Build GRCh38
rs917861182 Chr.3: 107378473-107378473 on Build GRCh38
rs748901462 Chr.3: 107378504-107378534 on Build GRCh38
rs746374757 Chr.3: 107378445-107378445 on Build GRCh38
rs112641427 Chr.3: 107378567-107378567 on Build GRCh38
rs183613011 Chr.3: 107378540-107378540 on Build GRCh38
rs748856786 Chr.3: 107378459-107378459 on Build GRCh38
rs751816868 Chr.3: 107378534-107378534 on Build GRCh38
rs758495479 Chr.3: 107378427-107378427 on Build GRCh38
rs778479927 Chr.3: 107378552-107378554 on Build GRCh38
rs752679485 Chr.3: 107378520-107378520 on Build GRCh38
rs778087401 Chr.3: 107378441-107378441 on Build GRCh38
rs992531976 Chr.3: 107378418-107378418 on Build GRCh38
rs749035116 Chr.3: 107378566-107378566 on Build GRCh38
rs962060760 Chr.3: 107378408-107378408 on Build GRCh38
rs771847715 Chr.3: 107378490-107378490 on Build GRCh38
rs760580137 Chr.3: 107378500-107378500 on Build GRCh38
rs760669969 Chr.3: 107378400-107378400 on Build GRCh38
rs766358872 Chr.3: 107378518-107378518 on Build GRCh38
rs770540209 Chr.3: 107378545-107378546 on Build GRCh38
rs748683794 Chr.3: 107378562-107378562 on Build GRCh38
rs761630856 Chr.3: 107378390-107378390 on Build GRCh38
rs750376409 Chr.3: 107378394-107378394 on Build GRCh38
rs764163308 Chr.3: 107378533-107378533 on Build GRCh38
rs142934626 Chr.3: 107378560-107378560 on Build GRCh38
rs201547203 Chr.3: 107378446-107378446 on Build GRCh38
rs146912601 Chr.3: 107378532-107378532 on Build GRCh38
rs752867905 Chr.3: 107378417-107378417 on Build GRCh38
rs767378185 Chr.3: 107378392-107378392 on Build GRCh38
rs779409946 Chr.3: 107378453-107378453 on Build GRCh38
rs3811061 Chr.3: 107378499-107378499 on Build GRCh38
rs372406570 Chr.3: 107378558-107378558 on Build GRCh38
rs367560807 Chr.3: 107378477-107378477 on Build GRCh38
rs761540911 Chr.3: 107378482-107378482 on Build GRCh38
rs543104179 Chr.3: 107378543-107378543 on Build GRCh38
rs34430473 Chr.3: 107378389-107378389 on Build GRCh38
rs757621575 Chr.3: 107378537-107378537 on Build GRCh38

View Full Product Details