Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 100988511-100988784 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C10orf2
Entrez Gene ID
56652
Gene Name
chromosome 10 open reading frame 2
Gene Aliases
ATXN8, IOSCA, MTDPS7, PEO, PEO1, PEOA3, PRLTS5, SANDO, SCA8, TWINL
UniGene
Hs.22678
Gene Chromosome Location
Chr.10: 100987527-100994403 on Build GRCh38
Transcripts
NM_001163812.1 NM_001163813.1 NM_001163814.1 NM_021830.4 XM_011539975.2 XM_017016437.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
913994 c.353C>T p.A118V Chr.10: 100988563-100988563 on Build GRCh38
1739638 c.454G>A p.E152K Chr.10: 100988664-100988664 on Build GRCh38
356681 c.445G>C p.E149Q Chr.10: 100988655-100988655 on Build GRCh38
7268578 c.552G>A p.K184K Chr.10: 100988762-100988762 on Build GRCh38
6442625 c.508C>T p.Q170* Chr.10: 100988718-100988718 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778192277 Chr.10: 100988715-100988716 on Build GRCh38
rs201767220 Chr.10: 100988663-100988663 on Build GRCh38
rs863223916 Chr.10: 100988550-100988550 on Build GRCh38
rs775463083 Chr.10: 100988702-100988702 on Build GRCh38
rs781279648 Chr.10: 100988605-100988605 on Build GRCh38
rs749624036 Chr.10: 100988627-100988627 on Build GRCh38
rs754261446 Chr.10: 100988752-100988752 on Build GRCh38
rs752699549 Chr.10: 100988563-100988563 on Build GRCh38
rs942646754 Chr.10: 100988603-100988603 on Build GRCh38
rs765038479 Chr.10: 100988551-100988551 on Build GRCh38
rs764957225 Chr.10: 100988694-100988694 on Build GRCh38
rs770407597 Chr.10: 100988676-100988676 on Build GRCh38
rs748648084 Chr.10: 100988761-100988761 on Build GRCh38
rs756400035 Chr.10: 100988743-100988743 on Build GRCh38
rs1051839743 Chr.10: 100988717-100988717 on Build GRCh38
rs375179111 Chr.10: 100988745-100988745 on Build GRCh38
rs759253288 Chr.10: 100988537-100988537 on Build GRCh38
rs746066180 Chr.10: 100988607-100988607 on Build GRCh38
rs143139606 Chr.10: 100988591-100988591 on Build GRCh38
rs780446227 Chr.10: 100988616-100988616 on Build GRCh38
rs767589437 Chr.10: 100988732-100988732 on Build GRCh38
rs755275071 Chr.10: 100988758-100988758 on Build GRCh38
rs764143807 Chr.10: 100988725-100988725 on Build GRCh38
rs760224175 Chr.10: 100988664-100988664 on Build GRCh38
rs375005531 Chr.10: 100988614-100988614 on Build GRCh38
rs757306944 Chr.10: 100988595-100988595 on Build GRCh38
rs757316627 Chr.10: 100988730-100988730 on Build GRCh38
rs765058338 Chr.10: 100988709-100988709 on Build GRCh38
rs199810842 Chr.10: 100988736-100988736 on Build GRCh38
rs863223917 Chr.10: 100988548-100988548 on Build GRCh38
rs758456031 Chr.10: 100988585-100988585 on Build GRCh38
rs772961299 Chr.10: 100988610-100988610 on Build GRCh38
rs751479054 Chr.10: 100988727-100988727 on Build GRCh38
rs148234280 Chr.10: 100988594-100988594 on Build GRCh38
rs143938897 Chr.10: 100988644-100988644 on Build GRCh38
rs886037832 Chr.10: 100988543-100988543 on Build GRCh38
rs370756491 Chr.10: 100988713-100988713 on Build GRCh38
rs200405447 Chr.10: 100988673-100988673 on Build GRCh38
rs776177991 Chr.10: 100988678-100988678 on Build GRCh38
rs761120352 Chr.10: 100988740-100988740 on Build GRCh38
rs377397119 Chr.10: 100988671-100988671 on Build GRCh38
rs759176256 Chr.10: 100988689-100988689 on Build GRCh38

View Full Product Details