Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 3928006-3928233 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP2A3
Entrez Gene ID
489
Gene Name
ATPase sarcoplasmic/endoplasmic reticulum Ca2+ transporting 3
Gene Aliases
SERCA3
UniGene
Hs.513870
Gene Chromosome Location
Chr.17: 3923869-3964464 on Build GRCh38
Transcripts
NM_005173.3 NM_174953.2 NM_174954.2 NM_174955.2 NM_174956.2 NM_174957.2 NM_174958.2 XM_011523881.2 XM_011523882.2 XM_011523884.2 XM_011523885.1 XM_011523888.2 XM_011523889.1 XM_017024692.1 XM_017024693.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3516965 c.3068+7C>T p.? Chr.17: 3928233-3928233 on Build GRCh38
6294646 c.3082-4C>A p.? Chr.17: 3928042-3928042 on Build GRCh38
6294647 c.533-4C>A p.? Chr.17: 3928042-3928042 on Build GRCh38
3937373 c.3100G>A p.V1034I Chr.17: 3928020-3928020 on Build GRCh38
6499392 c.555C>T p.L185L Chr.17: 3928016-3928016 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs891775239 Chr.17: 3928150-3928150 on Build GRCh38
rs746434821 Chr.17: 3928027-3928027 on Build GRCh38
rs768809922 Chr.17: 3928074-3928074 on Build GRCh38
rs953345619 Chr.17: 3928087-3928087 on Build GRCh38
rs776898428 Chr.17: 3928079-3928079 on Build GRCh38
rs41283383 Chr.17: 3928136-3928136 on Build GRCh38
rs775597121 Chr.17: 3928051-3928051 on Build GRCh38
rs755740079 Chr.17: 3928201-3928201 on Build GRCh38
rs987814408 Chr.17: 3928105-3928105 on Build GRCh38
rs946041288 Chr.17: 3928156-3928156 on Build GRCh38
rs766202362 Chr.17: 3928085-3928085 on Build GRCh38
rs143238410 Chr.17: 3928057-3928057 on Build GRCh38
rs752466680 Chr.17: 3928127-3928127 on Build GRCh38
rs772478788 Chr.17: 3928029-3928029 on Build GRCh38
rs781246473 Chr.17: 3928027-3928027 on Build GRCh38
rs1043107067 Chr.17: 3928157-3928157 on Build GRCh38
rs762732134 Chr.17: 3928083-3928083 on Build GRCh38
rs904420624 Chr.17: 3928160-3928160 on Build GRCh38
rs774138222 Chr.17: 3928089-3928089 on Build GRCh38
rs764000208 Chr.17: 3928196-3928196 on Build GRCh38
rs73328990 Chr.17: 3928198-3928198 on Build GRCh38
rs373321113 Chr.17: 3928200-3928200 on Build GRCh38
rs759473302 Chr.17: 3928190-3928190 on Build GRCh38
rs534734255 Chr.17: 3928128-3928128 on Build GRCh38

View Full Product Details