Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 77469044-77469301 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AHSA1
Entrez Gene ID
10598
Gene Name
activator of Hsp90 ATPase activity 1
Gene Aliases
AHA1, C14orf3, hAha1, p38
UniGene
Hs.204041
Gene Chromosome Location
Chr.14: 77457877-77469472 on Build GRCh38
Transcripts
NM_001321441.1 NM_012111.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5705773 c.913C>T p.R305* Chr.14: 77469145-77469145 on Build GRCh38
2139452 c.947G>C p.R316P Chr.14: 77469179-77469179 on Build GRCh38
3754133 c.945G>A p.T315T Chr.14: 77469177-77469177 on Build GRCh38
2139453 c.1002C>T p.G334G Chr.14: 77469234-77469234 on Build GRCh38
7140366 c.877G>T p.D293Y Chr.14: 77469109-77469109 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs111662785 Chr.14: 77469234-77469234 on Build GRCh38
rs766778621 Chr.14: 77469179-77469179 on Build GRCh38
rs765692305 Chr.14: 77469259-77469280 on Build GRCh38
rs200120741 Chr.14: 77469173-77469173 on Build GRCh38
rs146176111 Chr.14: 77469239-77469239 on Build GRCh38
rs768285181 Chr.14: 77469255-77469255 on Build GRCh38
rs773248402 Chr.14: 77469268-77469268 on Build GRCh38
rs113506805 Chr.14: 77469075-77469075 on Build GRCh38
rs761658354 Chr.14: 77469118-77469118 on Build GRCh38
rs375708095 Chr.14: 77469072-77469072 on Build GRCh38
rs372535976 Chr.14: 77469136-77469136 on Build GRCh38
rs906977051 Chr.14: 77469208-77469208 on Build GRCh38
rs916795638 Chr.14: 77469186-77469186 on Build GRCh38
rs773728757 Chr.14: 77469170-77469170 on Build GRCh38
rs7250 Chr.14: 77469177-77469177 on Build GRCh38
rs146211975 Chr.14: 77469210-77469210 on Build GRCh38
rs748904923 Chr.14: 77469244-77469244 on Build GRCh38
rs778315658 Chr.14: 77469257-77469257 on Build GRCh38
rs756053898 Chr.14: 77469195-77469195 on Build GRCh38
rs141659647 Chr.14: 77469235-77469235 on Build GRCh38
rs948223291 Chr.14: 77469193-77469193 on Build GRCh38
rs368267453 Chr.14: 77469081-77469081 on Build GRCh38
rs761408208 Chr.14: 77469082-77469082 on Build GRCh38
rs769486247 Chr.14: 77469068-77469068 on Build GRCh38
rs755174037 Chr.14: 77469194-77469194 on Build GRCh38
rs201280812 Chr.14: 77469275-77469275 on Build GRCh38
rs200880647 Chr.14: 77469117-77469117 on Build GRCh38
rs147994825 Chr.14: 77469238-77469238 on Build GRCh38
rs923918665 Chr.14: 77469146-77469146 on Build GRCh38
rs748895490 Chr.14: 77469094-77469094 on Build GRCh38
rs772182123 Chr.14: 77469265-77469265 on Build GRCh38
rs754103577 Chr.14: 77469181-77469181 on Build GRCh38
rs758852569 Chr.14: 77469233-77469233 on Build GRCh38
rs944735367 Chr.14: 77469221-77469221 on Build GRCh38
rs528306774 Chr.14: 77469264-77469264 on Build GRCh38
rs572141212 Chr.14: 77469109-77469109 on Build GRCh38
rs775092976 Chr.14: 77469088-77469088 on Build GRCh38
rs752784351 Chr.14: 77469198-77469198 on Build GRCh38
rs956397849 Chr.14: 77469172-77469172 on Build GRCh38

View Full Product Details