Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 68023052-68023299 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX28
Entrez Gene ID
55794
Gene Name
DEAD-box helicase 28
Gene Aliases
MDDX28
UniGene
Hs.458313
Gene Chromosome Location
Chr.16: 68021274-68023867 on Build GRCh38
Transcripts
NM_018380.3 NM_001271762.1 NM_001271763.1 NM_017803.4

Gene Symbol
DUS2
Entrez Gene ID
54920
Gene Name
dihydrouridine synthase 2
Gene Aliases
DUS2L, SMM1, URLC8
UniGene
Hs.534460
Gene Chromosome Location
Chr.16: 68022944-68079323 on Build GRCh38
Transcripts
NM_018380.3 NM_001271762.1 NM_001271763.1 NM_017803.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4832588 c.36C>G p.L12L Chr.16: 68023167-68023167 on Build GRCh38
6641695 c.124C>A p.L42I Chr.16: 68023079-68023079 on Build GRCh38
4845633 c.27C>G p.L9L Chr.16: 68023176-68023176 on Build GRCh38
302035 c.93A>C p.E31D Chr.16: 68023110-68023110 on Build GRCh38
4062070 c.14G>A p.R5Q Chr.16: 68023189-68023189 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764317601 Chr.16: 68023071-68023071 on Build GRCh38
rs780044099 Chr.16: 68023091-68023091 on Build GRCh38
rs771286069 Chr.16: 68023110-68023110 on Build GRCh38
rs770235784 Chr.16: 68023128-68023128 on Build GRCh38
rs914659667 Chr.16: 68023260-68023260 on Build GRCh38
rs748974461 Chr.16: 68023094-68023094 on Build GRCh38
rs756620466 Chr.16: 68023219-68023219 on Build GRCh38
rs751265571 Chr.16: 68023213-68023213 on Build GRCh38
rs752435052 Chr.16: 68023209-68023209 on Build GRCh38
rs763323843 Chr.16: 68023147-68023147 on Build GRCh38
rs766176001 Chr.16: 68023169-68023169 on Build GRCh38
rs984502183 Chr.16: 68023237-68023237 on Build GRCh38
rs777366429 Chr.16: 68023212-68023212 on Build GRCh38
rs76902372 Chr.16: 68023255-68023255 on Build GRCh38
rs761962423 Chr.16: 68023167-68023167 on Build GRCh38
rs774679672 Chr.16: 68023246-68023246 on Build GRCh38
rs751620852 Chr.16: 68023160-68023160 on Build GRCh38
rs757982492 Chr.16: 68023178-68023178 on Build GRCh38
rs559505232 Chr.16: 68023258-68023258 on Build GRCh38
rs759779862 Chr.16: 68023123-68023123 on Build GRCh38
rs748448080 Chr.16: 68023247-68023247 on Build GRCh38
rs754694008 Chr.16: 68023171-68023171 on Build GRCh38
rs1050631435 Chr.16: 68023191-68023191 on Build GRCh38
rs781521203 Chr.16: 68023186-68023186 on Build GRCh38
rs141515984 Chr.16: 68023204-68023204 on Build GRCh38
rs775831161 Chr.16: 68023139-68023139 on Build GRCh38
rs200017416 Chr.16: 68023105-68023105 on Build GRCh38
rs756071106 Chr.16: 68023085-68023085 on Build GRCh38
rs1034206482 Chr.16: 68023228-68023228 on Build GRCh38
rs770507825 Chr.16: 68023185-68023185 on Build GRCh38
rs778385465 Chr.16: 68023101-68023101 on Build GRCh38
rs921173279 Chr.16: 68023079-68023079 on Build GRCh38
rs749618467 Chr.16: 68023233-68023233 on Build GRCh38
rs758213519 Chr.16: 68023211-68023211 on Build GRCh38
rs237831 Chr.16: 68023193-68023193 on Build GRCh38
rs747119709 Chr.16: 68023206-68023209 on Build GRCh38
rs752347194 Chr.16: 68023177-68023177 on Build GRCh38
rs769171202 Chr.16: 68023234-68023234 on Build GRCh38
rs150933726 Chr.16: 68023266-68023266 on Build GRCh38
rs746436723 Chr.16: 68023187-68023187 on Build GRCh38
rs764262500 Chr.16: 68023151-68023151 on Build GRCh38
rs750372765 Chr.16: 68023084-68023084 on Build GRCh38
rs746456268 Chr.16: 68023182-68023182 on Build GRCh38
rs764247926 Chr.16: 68023252-68023252 on Build GRCh38
rs780811590 Chr.16: 68023214-68023214 on Build GRCh38
rs562009202 Chr.16: 68023190-68023190 on Build GRCh38
rs923429737 Chr.16: 68023174-68023174 on Build GRCh38
rs148388647 Chr.16: 68023201-68023201 on Build GRCh38
rs1001410260 Chr.16: 68023215-68023215 on Build GRCh38
rs746529299 Chr.16: 68023109-68023109 on Build GRCh38
rs780718607 Chr.16: 68023229-68023229 on Build GRCh38
rs778740776 Chr.16: 68023176-68023176 on Build GRCh38
rs777420050 Chr.16: 68023180-68023180 on Build GRCh38
rs774734563 Chr.16: 68023197-68023197 on Build GRCh38
rs923440272 Chr.16: 68023261-68023261 on Build GRCh38
rs765166334 Chr.16: 68023207-68023207 on Build GRCh38
rs200362568 Chr.16: 68023173-68023173 on Build GRCh38
rs754821676 Chr.16: 68023097-68023097 on Build GRCh38
rs201938424 Chr.16: 68023198-68023198 on Build GRCh38

View Full Product Details