Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 99430373-99430646 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GOT1
Entrez Gene ID
2805
Gene Name
glutamic-oxaloacetic transaminase 1
Gene Aliases
AST1, ASTQTL1, GIG18, cAspAT, cCAT
UniGene
Hs.500756
Gene Chromosome Location
Chr.10: 99396870-99430773 on Build GRCh38
Transcripts
NM_002079.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5531784 c.48C>T p.V16V Chr.10: 99430518-99430518 on Build GRCh38
682540 c.63C>A p.L21L Chr.10: 99430503-99430503 on Build GRCh38
6307845 c.61C>A p.L21I Chr.10: 99430505-99430505 on Build GRCh38
6861689 c.77G>A p.R26K Chr.10: 99430489-99430489 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs375375767 Chr.10: 99430600-99430600 on Build GRCh38
rs373418737 Chr.10: 99430452-99430452 on Build GRCh38
rs779521968 Chr.10: 99430428-99430428 on Build GRCh38
rs774104270 Chr.10: 99430611-99430611 on Build GRCh38
rs774226921 Chr.10: 99430476-99430476 on Build GRCh38
rs768475860 Chr.10: 99430473-99430473 on Build GRCh38
rs909656692 Chr.10: 99430471-99430471 on Build GRCh38
rs745448004 Chr.10: 99430451-99430451 on Build GRCh38
rs767534084 Chr.10: 99430506-99430506 on Build GRCh38
rs79694820 Chr.10: 99430392-99430392 on Build GRCh38
rs1025302991 Chr.10: 99430621-99430621 on Build GRCh38
rs903247664 Chr.10: 99430484-99430484 on Build GRCh38
rs755345377 Chr.10: 99430554-99430554 on Build GRCh38
rs756261660 Chr.10: 99430411-99430411 on Build GRCh38
rs769347810 Chr.10: 99430599-99430599 on Build GRCh38
rs761665544 Chr.10: 99430616-99430616 on Build GRCh38
rs1039583210 Chr.10: 99430414-99430414 on Build GRCh38
rs11557841 Chr.10: 99430525-99430525 on Build GRCh38
rs750467322 Chr.10: 99430526-99430526 on Build GRCh38
rs374880822 Chr.10: 99430605-99430605 on Build GRCh38
rs770282613 Chr.10: 99430587-99430587 on Build GRCh38
rs762666294 Chr.10: 99430463-99430463 on Build GRCh38
rs754094707 Chr.10: 99430545-99430545 on Build GRCh38
rs2234894 Chr.10: 99430529-99430529 on Build GRCh38
rs760867860 Chr.10: 99430540-99430540 on Build GRCh38
rs901102068 Chr.10: 99430436-99430436 on Build GRCh38
rs202105295 Chr.10: 99430419-99430419 on Build GRCh38
rs1000322922 Chr.10: 99430496-99430496 on Build GRCh38
rs772511514 Chr.10: 99430572-99430572 on Build GRCh38
rs563842353 Chr.10: 99430628-99430628 on Build GRCh38
rs866444380 Chr.10: 99430393-99430393 on Build GRCh38
rs748965656 Chr.10: 99430604-99430604 on Build GRCh38
rs755408557 Chr.10: 99430427-99430427 on Build GRCh38
rs532815687 Chr.10: 99430400-99430400 on Build GRCh38
rs778400032 Chr.10: 99430590-99430590 on Build GRCh38
rs779398030 Chr.10: 99430562-99430562 on Build GRCh38
rs766454263 Chr.10: 99430542-99430542 on Build GRCh38
rs748686612 Chr.10: 99430437-99430437 on Build GRCh38
rs758966981 Chr.10: 99430578-99430578 on Build GRCh38
rs772753323 Chr.10: 99430442-99430442 on Build GRCh38
rs759679111 Chr.10: 99430623-99430623 on Build GRCh38
rs761720482 Chr.10: 99430499-99430499 on Build GRCh38
rs749604649 Chr.10: 99430425-99430425 on Build GRCh38
rs745311133 Chr.10: 99430592-99430592 on Build GRCh38
rs753151095 Chr.10: 99430570-99430570 on Build GRCh38
rs867790769 Chr.10: 99430498-99430498 on Build GRCh38
rs1024531946 Chr.10: 99430432-99430432 on Build GRCh38

View Full Product Details