Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 32372085-32372314 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASXL1
Entrez Gene ID
171023
Gene Name
additional sex combs like 1, transcriptional regulator
Gene Aliases
BOPS, MDS
UniGene
Hs.374043
Gene Chromosome Location
Chr.20: 32358062-32439319 on Build GRCh38
Transcripts
NM_001164603.1 NM_015338.5 XM_006723727.3 XM_006723728.3 XM_006723730.3 XM_006723732.2 XM_011528648.2 XM_011528652.2 XM_017027704.1 XM_017027705.1 XM_017027706.1

SNP Detail

SNP ID Chromosome Location
rs757417637 Chr.20: 32372116-32372116 on Build GRCh38
rs968681105 Chr.20: 32372161-32372161 on Build GRCh38
rs745897173 Chr.20: 32372186-32372186 on Build GRCh38
rs1005292129 Chr.20: 32372125-32372125 on Build GRCh38
rs971246274 Chr.20: 32372216-32372216 on Build GRCh38
rs557659733 Chr.20: 32372119-32372119 on Build GRCh38
rs927124359 Chr.20: 32372230-32372230 on Build GRCh38
rs371489431 Chr.20: 32372112-32372112 on Build GRCh38
rs769925859 Chr.20: 32372199-32372199 on Build GRCh38
rs374193788 Chr.20: 32372224-32372224 on Build GRCh38
rs781128104 Chr.20: 32372176-32372176 on Build GRCh38
rs776622622 Chr.20: 32372284-32372284 on Build GRCh38
rs765792159 Chr.20: 32372240-32372240 on Build GRCh38
rs559005193 Chr.20: 32372109-32372109 on Build GRCh38
rs994207526 Chr.20: 32372187-32372187 on Build GRCh38

View Full Product Details