Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 49378387-49378660 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C16orf78
Entrez Gene ID
123970
Gene Name
chromosome 16 open reading frame 78
Gene Aliases
-
UniGene
Hs.125875
Gene Chromosome Location
Chr.16: 49373808-49399431 on Build GRCh38
Transcripts
NM_144602.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2833528 c.339G>T p.G113G Chr.16: 49378538-49378538 on Build GRCh38
971127 c.301G>A p.A101T Chr.16: 49378500-49378500 on Build GRCh38
2833527 c.338G>A p.G113E Chr.16: 49378537-49378537 on Build GRCh38
4189042 c.372C>G p.I124M Chr.16: 49378571-49378571 on Build GRCh38
6614579 c.318C>A p.L106L Chr.16: 49378517-49378517 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs952400148 Chr.16: 49378622-49378622 on Build GRCh38
rs763184906 Chr.16: 49378443-49378443 on Build GRCh38
rs374161160 Chr.16: 49378424-49378424 on Build GRCh38
rs140023005 Chr.16: 49378444-49378444 on Build GRCh38
rs150674263 Chr.16: 49378542-49378542 on Build GRCh38
rs775578028 Chr.16: 49378519-49378519 on Build GRCh38
rs767886935 Chr.16: 49378537-49378537 on Build GRCh38
rs78114813 Chr.16: 49378561-49378561 on Build GRCh38
rs548363863 Chr.16: 49378545-49378545 on Build GRCh38
rs764434405 Chr.16: 49378529-49378529 on Build GRCh38
rs865863717 Chr.16: 49378469-49378469 on Build GRCh38
rs143423404 Chr.16: 49378496-49378496 on Build GRCh38
rs780359681 Chr.16: 49378559-49378559 on Build GRCh38
rs201013677 Chr.16: 49378574-49378574 on Build GRCh38
rs74431342 Chr.16: 49378510-49378510 on Build GRCh38
rs193149573 Chr.16: 49378631-49378631 on Build GRCh38
rs189028366 Chr.16: 49378420-49378420 on Build GRCh38
rs758114215 Chr.16: 49378546-49378546 on Build GRCh38
rs774682583 Chr.16: 49378616-49378616 on Build GRCh38
rs1020101242 Chr.16: 49378531-49378531 on Build GRCh38
rs891789829 Chr.16: 49378522-49378522 on Build GRCh38
rs938955852 Chr.16: 49378412-49378412 on Build GRCh38
rs747758290 Chr.16: 49378497-49378497 on Build GRCh38
rs368310360 Chr.16: 49378466-49378466 on Build GRCh38
rs752090725 Chr.16: 49378625-49378625 on Build GRCh38
rs941609165 Chr.16: 49378421-49378421 on Build GRCh38
rs762276523 Chr.16: 49378534-49378534 on Build GRCh38
rs550915813 Chr.16: 49378586-49378586 on Build GRCh38
rs768727180 Chr.16: 49378615-49378615 on Build GRCh38
rs769640378 Chr.16: 49378562-49378562 on Build GRCh38
rs868089141 Chr.16: 49378465-49378465 on Build GRCh38
rs27813 Chr.16: 49378620-49378620 on Build GRCh38
rs781755813 Chr.16: 49378472-49378472 on Build GRCh38
rs774574783 Chr.16: 49378530-49378530 on Build GRCh38
rs751967122 Chr.16: 49378459-49378459 on Build GRCh38
rs931007479 Chr.16: 49378450-49378450 on Build GRCh38
rs145444097 Chr.16: 49378509-49378509 on Build GRCh38
rs775533178 Chr.16: 49378591-49378591 on Build GRCh38
rs373637026 Chr.16: 49378500-49378500 on Build GRCh38
rs780874323 Chr.16: 49378569-49378569 on Build GRCh38
rs749436378 Chr.16: 49378600-49378600 on Build GRCh38
rs758124337 Chr.16: 49378499-49378499 on Build GRCh38
rs753255711 Chr.16: 49378473-49378473 on Build GRCh38
rs1047205354 Chr.16: 49378411-49378411 on Build GRCh38
rs763094885 Chr.16: 49378523-49378523 on Build GRCh38
rs757448859 Chr.16: 49378612-49378620 on Build GRCh38
rs547715915 Chr.16: 49378579-49378579 on Build GRCh38
rs138952034 Chr.16: 49378543-49378543 on Build GRCh38
rs757691339 Chr.16: 49378468-49378468 on Build GRCh38
rs745926781 Chr.16: 49378515-49378515 on Build GRCh38
rs202090196 Chr.16: 49378451-49378451 on Build GRCh38
rs759225468 Chr.16: 49378610-49378612 on Build GRCh38
rs767880404 Chr.16: 49378632-49378632 on Build GRCh38
rs376632421 Chr.16: 49378575-49378575 on Build GRCh38
rs752966811 Chr.16: 49378442-49378442 on Build GRCh38
rs372064943 Chr.16: 49378491-49378491 on Build GRCh38
rs765415827 Chr.16: 49378437-49378437 on Build GRCh38

View Full Product Details