Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 58470512-58470777 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APCDD1L
Entrez Gene ID
164284
Gene Name
APC down-regulated 1 like
Gene Aliases
-
UniGene
Hs.119286
Gene Chromosome Location
Chr.20: 58459101-58514938 on Build GRCh38
Transcripts
NM_001304787.1 NM_153360.2 NR_130908.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
238810 c.160C>T p.R54C Chr.20: 58470637-58470637 on Build GRCh38
7226558 c.121T>C p.C41R Chr.20: 58470709-58470709 on Build GRCh38
4783649 c.219A>G p.T73T Chr.20: 58470611-58470611 on Build GRCh38
73693 c.148A>G p.I50V Chr.20: 58470649-58470649 on Build GRCh38
1751515 c.186A>G p.T62T Chr.20: 58470611-58470611 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199788439 Chr.20: 58470637-58470637 on Build GRCh38
rs3946003 Chr.20: 58470709-58470709 on Build GRCh38
rs747250193 Chr.20: 58470652-58470652 on Build GRCh38
rs142502149 Chr.20: 58470701-58470701 on Build GRCh38
rs772487718 Chr.20: 58470650-58470650 on Build GRCh38
rs751797316 Chr.20: 58470714-58470714 on Build GRCh38
rs199697401 Chr.20: 58470670-58470670 on Build GRCh38
rs766383390 Chr.20: 58470585-58470585 on Build GRCh38
rs1048417652 Chr.20: 58470644-58470644 on Build GRCh38
rs561798456 Chr.20: 58470706-58470706 on Build GRCh38
rs140187713 Chr.20: 58470708-58470708 on Build GRCh38
rs147147447 Chr.20: 58470703-58470703 on Build GRCh38
rs557310543 Chr.20: 58470546-58470546 on Build GRCh38
rs1039659331 Chr.20: 58470577-58470577 on Build GRCh38
rs1980576 Chr.20: 58470611-58470611 on Build GRCh38
rs750968695 Chr.20: 58470714-58470714 on Build GRCh38
rs995154561 Chr.20: 58470629-58470629 on Build GRCh38
rs888411625 Chr.20: 58470679-58470679 on Build GRCh38
rs866961837 Chr.20: 58470620-58470620 on Build GRCh38
rs188971133 Chr.20: 58470612-58470612 on Build GRCh38
rs746300653 Chr.20: 58470647-58470647 on Build GRCh38
rs113379816 Chr.20: 58470740-58470740 on Build GRCh38
rs773610872 Chr.20: 58470742-58470742 on Build GRCh38
rs780236950 Chr.20: 58470651-58470651 on Build GRCh38
rs768750681 Chr.20: 58470568-58470568 on Build GRCh38
rs867499364 Chr.20: 58470557-58470557 on Build GRCh38
rs372663041 Chr.20: 58470638-58470638 on Build GRCh38
rs779530434 Chr.20: 58470640-58470640 on Build GRCh38
rs751449964 Chr.20: 58470594-58470594 on Build GRCh38
rs148393493 Chr.20: 58470636-58470636 on Build GRCh38
rs575647147 Chr.20: 58470598-58470598 on Build GRCh38
rs535618918 Chr.20: 58470545-58470545 on Build GRCh38
rs768919971 Chr.20: 58470656-58470656 on Build GRCh38
rs112628550 Chr.20: 58470735-58470735 on Build GRCh38
rs763906482 Chr.20: 58470705-58470705 on Build GRCh38
rs776806093 Chr.20: 58470668-58470668 on Build GRCh38
rs375466344 Chr.20: 58470702-58470702 on Build GRCh38
rs866877267 Chr.20: 58470634-58470634 on Build GRCh38
rs371170776 Chr.20: 58470563-58470563 on Build GRCh38
rs780513473 Chr.20: 58470723-58470723 on Build GRCh38
rs573179097 Chr.20: 58470672-58470672 on Build GRCh38
rs562721222 Chr.20: 58470718-58470718 on Build GRCh38
rs769752179 Chr.20: 58470671-58470671 on Build GRCh38
rs776646617 Chr.20: 58470573-58470573 on Build GRCh38
rs761861941 Chr.20: 58470583-58470583 on Build GRCh38
rs752403111 Chr.20: 58470630-58470630 on Build GRCh38

View Full Product Details