Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 92354087-92354344 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAT3
Entrez Gene ID
120114
Gene Name
FAT atypical cadherin 3
Gene Aliases
CDHF15, CDHR10, hFat3
UniGene
Hs.98523
Gene Chromosome Location
Chr.11: 92224636-92896473 on Build GRCh38
Transcripts
NM_001008781.2 XM_017017178.1 XM_017017179.1 XM_017017180.1 XM_017017181.1 XM_017017182.1 XM_017017183.1 XM_017017184.1 XM_017017186.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3870411 c.1975G>A p.E659K Chr.11: 92354087-92354087 on Build GRCh38
2223301 c.2188C>T p.P730S Chr.11: 92354300-92354300 on Build GRCh38
2223271 c.2009C>T p.S670L Chr.11: 92354121-92354121 on Build GRCh38
4683577 c.2137C>A p.L713I Chr.11: 92354249-92354249 on Build GRCh38
5144332 c.2104G>T p.A702S Chr.11: 92354216-92354216 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767626429 Chr.11: 92354283-92354283 on Build GRCh38
rs778595053 Chr.11: 92354210-92354210 on Build GRCh38
rs781607497 Chr.11: 92354230-92354230 on Build GRCh38
rs377153421 Chr.11: 92354132-92354132 on Build GRCh38
rs754627027 Chr.11: 92354204-92354204 on Build GRCh38
rs770182957 Chr.11: 92354138-92354138 on Build GRCh38
rs755926119 Chr.11: 92354189-92354189 on Build GRCh38
rs370396712 Chr.11: 92354234-92354234 on Build GRCh38
rs775755926 Chr.11: 92354146-92354146 on Build GRCh38
rs267603241 Chr.11: 92354310-92354310 on Build GRCh38
rs536693987 Chr.11: 92354307-92354307 on Build GRCh38
rs374844397 Chr.11: 92354252-92354252 on Build GRCh38
rs926621887 Chr.11: 92354293-92354293 on Build GRCh38
rs770185976 Chr.11: 92354242-92354242 on Build GRCh38
rs766241873 Chr.11: 92354200-92354200 on Build GRCh38
rs957827238 Chr.11: 92354227-92354227 on Build GRCh38
rs1049371717 Chr.11: 92354205-92354205 on Build GRCh38
rs753590352 Chr.11: 92354201-92354201 on Build GRCh38
rs757795933 Chr.11: 92354220-92354220 on Build GRCh38
rs567803449 Chr.11: 92354272-92354272 on Build GRCh38
rs761744347 Chr.11: 92354279-92354279 on Build GRCh38
rs1040386421 Chr.11: 92354233-92354233 on Build GRCh38
rs982502822 Chr.11: 92354266-92354266 on Build GRCh38
rs377761938 Chr.11: 92354264-92354264 on Build GRCh38
rs761843855 Chr.11: 92354171-92354171 on Build GRCh38
rs750357297 Chr.11: 92354184-92354184 on Build GRCh38
rs753644851 Chr.11: 92354320-92354320 on Build GRCh38
rs771300455 Chr.11: 92354127-92354127 on Build GRCh38
rs140299757 Chr.11: 92354256-92354256 on Build GRCh38
rs373882316 Chr.11: 92354245-92354245 on Build GRCh38
rs372914793 Chr.11: 92354109-92354109 on Build GRCh38
rs746162335 Chr.11: 92354137-92354137 on Build GRCh38
rs764210894 Chr.11: 92354162-92354162 on Build GRCh38
rs774369902 Chr.11: 92354167-92354167 on Build GRCh38
rs747545363 Chr.11: 92354211-92354211 on Build GRCh38
rs35515591 Chr.11: 92354319-92354320 on Build GRCh38
rs747540079 Chr.11: 92354125-92354125 on Build GRCh38
rs767293755 Chr.11: 92354172-92354172 on Build GRCh38
rs763131522 Chr.11: 92354153-92354153 on Build GRCh38
rs773058850 Chr.11: 92354306-92354306 on Build GRCh38

View Full Product Details