Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 158859112-158859337 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FNIP2
Entrez Gene ID
57600
Gene Name
folliculin interacting protein 2
Gene Aliases
FNIPL, MAPO1
UniGene
Hs.652441
Gene Chromosome Location
Chr.4: 158768995-158908050 on Build GRCh38
Transcripts
NM_001323916.1 NM_020840.2 XM_005263156.1 XM_005263158.1 XM_005263160.3 XM_011532154.2 XM_017008487.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3601478 c.953C>T p.S318F Chr.4: 158859152-158859152 on Build GRCh38
4950566 c.1000T>A p.F334I Chr.4: 158859199-158859199 on Build GRCh38
5669698 c.966A>T p.K322N Chr.4: 158859165-158859165 on Build GRCh38
1186687 c.1045A>G p.S349G Chr.4: 158859244-158859244 on Build GRCh38
5160769 c.1059+8G>A p.? Chr.4: 158859266-158859266 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759571389 Chr.4: 158859192-158859194 on Build GRCh38
rs368406944 Chr.4: 158859194-158859194 on Build GRCh38
rs756439134 Chr.4: 158859283-158859283 on Build GRCh38
rs35074996 Chr.4: 158859214-158859215 on Build GRCh38
rs768614234 Chr.4: 158859154-158859154 on Build GRCh38
rs990054675 Chr.4: 158859202-158859202 on Build GRCh38
rs772951747 Chr.4: 158859210-158859210 on Build GRCh38
rs1028761895 Chr.4: 158859148-158859148 on Build GRCh38
rs754018559 Chr.4: 158859286-158859286 on Build GRCh38
rs569434783 Chr.4: 158859145-158859145 on Build GRCh38
rs761604559 Chr.4: 158859190-158859190 on Build GRCh38
rs879422631 Chr.4: 158859225-158859225 on Build GRCh38
rs766030489 Chr.4: 158859243-158859243 on Build GRCh38
rs371941985 Chr.4: 158859245-158859245 on Build GRCh38
rs762588110 Chr.4: 158859217-158859217 on Build GRCh38
rs556675960 Chr.4: 158859153-158859153 on Build GRCh38
rs987295880 Chr.4: 158859170-158859170 on Build GRCh38
rs536744472 Chr.4: 158859147-158859147 on Build GRCh38
rs768062132 Chr.4: 158859268-158859268 on Build GRCh38
rs375042093 Chr.4: 158859277-158859277 on Build GRCh38
rs967358022 Chr.4: 158859236-158859236 on Build GRCh38
rs776345131 Chr.4: 158859156-158859156 on Build GRCh38
rs552453601 Chr.4: 158859144-158859144 on Build GRCh38
rs978341381 Chr.4: 158859310-158859310 on Build GRCh38
rs532853556 Chr.4: 158859142-158859142 on Build GRCh38
rs573821947 Chr.4: 158859284-158859284 on Build GRCh38
rs760219138 Chr.4: 158859197-158859197 on Build GRCh38
rs755498242 Chr.4: 158859255-158859255 on Build GRCh38
rs915449830 Chr.4: 158859213-158859213 on Build GRCh38

View Full Product Details