Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 87156482-87156755 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C9orf170
Entrez Gene ID
401535
Gene Name
chromosome 9 open reading frame 170
Gene Aliases
-
UniGene
Hs.657740
Gene Chromosome Location
Chr.9: 87148644-87159726 on Build GRCh38
Transcripts
NM_001001709.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4410407 c.248G>C p.G83A Chr.9: 87156652-87156652 on Build GRCh38
4375607 c.279G>C p.L93F Chr.9: 87156683-87156683 on Build GRCh38
5998452 c.329G>C p.R110T Chr.9: 87156733-87156733 on Build GRCh38
1110780 c.331A>C p.R111R Chr.9: 87156735-87156735 on Build GRCh38
4171901 c.181C>T p.R61W Chr.9: 87156585-87156585 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758651445 Chr.9: 87156668-87156668 on Build GRCh38
rs371500498 Chr.9: 87156531-87156531 on Build GRCh38
rs139907209 Chr.9: 87156578-87156578 on Build GRCh38
rs968554326 Chr.9: 87156548-87156548 on Build GRCh38
rs539508145 Chr.9: 87156696-87156696 on Build GRCh38
rs775002142 Chr.9: 87156729-87156729 on Build GRCh38
rs79710047 Chr.9: 87156681-87156681 on Build GRCh38
rs776271627 Chr.9: 87156728-87156728 on Build GRCh38
rs771815948 Chr.9: 87156715-87156715 on Build GRCh38
rs758499354 Chr.9: 87156571-87156571 on Build GRCh38
rs180970139 Chr.9: 87156569-87156569 on Build GRCh38
rs777739838 Chr.9: 87156685-87156685 on Build GRCh38
rs111327086 Chr.9: 87156673-87156673 on Build GRCh38
rs780078126 Chr.9: 87156670-87156670 on Build GRCh38
rs142866011 Chr.9: 87156586-87156586 on Build GRCh38
rs916882701 Chr.9: 87156610-87156610 on Build GRCh38
rs892445409 Chr.9: 87156688-87156688 on Build GRCh38
rs151075470 Chr.9: 87156650-87156650 on Build GRCh38
rs371880236 Chr.9: 87156656-87156656 on Build GRCh38
rs749248786 Chr.9: 87156692-87156692 on Build GRCh38
rs992739544 Chr.9: 87156590-87156590 on Build GRCh38
rs532166941 Chr.9: 87156551-87156551 on Build GRCh38
rs773688800 Chr.9: 87156623-87156623 on Build GRCh38
rs761055290 Chr.9: 87156585-87156585 on Build GRCh38
rs765777241 Chr.9: 87156720-87156731 on Build GRCh38
rs376327560 Chr.9: 87156666-87156666 on Build GRCh38
rs1034088072 Chr.9: 87156690-87156690 on Build GRCh38
rs745311825 Chr.9: 87156703-87156703 on Build GRCh38
rs748861395 Chr.9: 87156617-87156617 on Build GRCh38
rs374885744 Chr.9: 87156697-87156697 on Build GRCh38
rs570811141 Chr.9: 87156555-87156555 on Build GRCh38
rs367976208 Chr.9: 87156622-87156622 on Build GRCh38
rs753331140 Chr.9: 87156657-87156657 on Build GRCh38
rs547980373 Chr.9: 87156554-87156554 on Build GRCh38
rs781071295 Chr.9: 87156572-87156572 on Build GRCh38
rs76452933 Chr.9: 87156654-87156654 on Build GRCh38
rs779316341 Chr.9: 87156559-87156559 on Build GRCh38
rs767015589 Chr.9: 87156645-87156645 on Build GRCh38
rs765379868 Chr.9: 87156661-87156661 on Build GRCh38
rs113607905 Chr.9: 87156701-87156701 on Build GRCh38
rs186548506 Chr.9: 87156730-87156730 on Build GRCh38
rs533463954 Chr.9: 87156557-87156557 on Build GRCh38
rs761068316 Chr.9: 87156529-87156529 on Build GRCh38
rs759268715 Chr.9: 87156638-87156638 on Build GRCh38

View Full Product Details