Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 30663509-30663756 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUSP18
Entrez Gene ID
150290
Gene Name
dual specificity phosphatase 18
Gene Aliases
DSP18, DUSP20, LMWDSP20
UniGene
Hs.517544
Gene Chromosome Location
Chr.22: 30652051-30667890 on Build GRCh38
Transcripts
NM_001304794.1 NM_001304795.1 NM_001304796.1 NM_152511.4 XM_005261368.4 XM_006724148.3 XM_011529920.2 XM_011529921.2 XM_017028627.1 XM_017028628.1 NM_001318371.1

Gene Symbol
SLC35E4
Entrez Gene ID
339665
Gene Name
solute carrier family 35 member E4
Gene Aliases
-
UniGene
Hs.660384
Gene Chromosome Location
Chr.22: 30634148-30669016 on Build GRCh38
Transcripts
NM_001304794.1 NM_001304795.1 NM_001304796.1 NM_152511.4 XM_005261368.4 XM_006724148.3 XM_011529920.2 XM_011529921.2 XM_017028627.1 XM_017028628.1 NM_001318371.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2937363 c.388G>A p.A130T Chr.22: 30663616-30663616 on Build GRCh38
2937364 c.370G>A p.A124T Chr.22: 30663634-30663634 on Build GRCh38
4679713 c.277G>A p.V93M Chr.22: 30663727-30663727 on Build GRCh38
1415744 c.387C>T p.D129D Chr.22: 30663617-30663617 on Build GRCh38
5707632 c.435C>A p.S145R Chr.22: 30663569-30663569 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757521320 Chr.22: 30663616-30663616 on Build GRCh38
rs775345868 Chr.22: 30663568-30663568 on Build GRCh38
rs769777449 Chr.22: 30663627-30663627 on Build GRCh38
rs755916451 Chr.22: 30663697-30663697 on Build GRCh38
rs201450102 Chr.22: 30663676-30663676 on Build GRCh38
rs753195199 Chr.22: 30663666-30663666 on Build GRCh38
rs377209045 Chr.22: 30663644-30663644 on Build GRCh38
rs749210779 Chr.22: 30663700-30663700 on Build GRCh38
rs368849519 Chr.22: 30663656-30663656 on Build GRCh38
rs577122514 Chr.22: 30663604-30663604 on Build GRCh38
rs768947169 Chr.22: 30663702-30663702 on Build GRCh38
rs1023241324 Chr.22: 30663720-30663720 on Build GRCh38
rs761476552 Chr.22: 30663584-30663584 on Build GRCh38
rs916368244 Chr.22: 30663552-30663552 on Build GRCh38
rs372836051 Chr.22: 30663609-30663609 on Build GRCh38
rs140097338 Chr.22: 30663728-30663728 on Build GRCh38
rs765722710 Chr.22: 30663600-30663600 on Build GRCh38
rs757467853 Chr.22: 30663682-30663682 on Build GRCh38
rs540794868 Chr.22: 30663662-30663662 on Build GRCh38
rs572091331 Chr.22: 30663635-30663635 on Build GRCh38
rs889237064 Chr.22: 30663686-30663686 on Build GRCh38
rs201513667 Chr.22: 30663543-30663543 on Build GRCh38
rs935279660 Chr.22: 30663615-30663615 on Build GRCh38
rs763517501 Chr.22: 30663580-30663580 on Build GRCh38
rs750679648 Chr.22: 30663695-30663695 on Build GRCh38
rs202138261 Chr.22: 30663675-30663675 on Build GRCh38
rs754173853 Chr.22: 30663592-30663592 on Build GRCh38
rs200346696 Chr.22: 30663708-30663708 on Build GRCh38
rs747647004 Chr.22: 30663706-30663706 on Build GRCh38
rs767138750 Chr.22: 30663655-30663655 on Build GRCh38
rs148574866 Chr.22: 30663608-30663608 on Build GRCh38
rs146604795 Chr.22: 30663699-30663699 on Build GRCh38
rs775683131 Chr.22: 30663730-30663730 on Build GRCh38
rs751667708 Chr.22: 30663678-30663678 on Build GRCh38
rs775433384 Chr.22: 30663634-30663634 on Build GRCh38
rs773998729 Chr.22: 30663583-30663583 on Build GRCh38
rs768164376 Chr.22: 30663637-30663637 on Build GRCh38
rs144452811 Chr.22: 30663665-30663665 on Build GRCh38
rs144847527 Chr.22: 30663569-30663569 on Build GRCh38
rs760340746 Chr.22: 30663727-30663727 on Build GRCh38
rs753147447 Chr.22: 30663602-30663602 on Build GRCh38
rs368354166 Chr.22: 30663591-30663591 on Build GRCh38
rs149449436 Chr.22: 30663557-30663557 on Build GRCh38
rs773766290 Chr.22: 30663641-30663641 on Build GRCh38
rs755387475 Chr.22: 30663594-30663594 on Build GRCh38
rs766693364 Chr.22: 30663587-30663587 on Build GRCh38
rs781613794 Chr.22: 30663617-30663617 on Build GRCh38
rs772790959 Chr.22: 30663709-30663709 on Build GRCh38
rs745644005 Chr.22: 30663623-30663623 on Build GRCh38
rs781558841 Chr.22: 30663694-30663694 on Build GRCh38

View Full Product Details