Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 32029657-32029900 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCM2L
Entrez Gene ID
140706
Gene Name
CCM2 like scaffolding protein
Gene Aliases
C20orf160, dJ310O13.5
UniGene
Hs.382151
Gene Chromosome Location
Chr.20: 32010379-32032182 on Build GRCh38
Transcripts
NM_080625.3 XM_006723706.3 XM_011528566.1 XM_011528567.2 XM_011528568.2 XM_011528569.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6622894 c.1281G>A p.R427R Chr.20: 32029781-32029781 on Build GRCh38
2888952 c.1385G>A p.R462H Chr.20: 32029821-32029821 on Build GRCh38
2888946 c.1288G>A p.E430K Chr.20: 32029724-32029724 on Build GRCh38
4445589 c.1392C>T p.F464F Chr.20: 32029828-32029828 on Build GRCh38
6622892 c.1240C>T p.R414* Chr.20: 32029740-32029740 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770774407 Chr.20: 32029849-32029849 on Build GRCh38
rs752352558 Chr.20: 32029741-32029741 on Build GRCh38
rs760516738 Chr.20: 32029792-32029792 on Build GRCh38
rs772023821 Chr.20: 32029846-32029846 on Build GRCh38
rs201862826 Chr.20: 32029769-32029769 on Build GRCh38
rs764356668 Chr.20: 32029739-32029739 on Build GRCh38
rs201292997 Chr.20: 32029810-32029810 on Build GRCh38
rs535524553 Chr.20: 32029680-32029680 on Build GRCh38
rs761982941 Chr.20: 32029855-32029855 on Build GRCh38
rs540781252 Chr.20: 32029751-32029751 on Build GRCh38
rs764802834 Chr.20: 32029724-32029724 on Build GRCh38
rs878977010 Chr.20: 32029829-32029829 on Build GRCh38
rs760422829 Chr.20: 32029696-32029696 on Build GRCh38
rs766134324 Chr.20: 32029795-32029795 on Build GRCh38
rs774476140 Chr.20: 32029685-32029685 on Build GRCh38
rs939007875 Chr.20: 32029716-32029716 on Build GRCh38
rs748282429 Chr.20: 32029844-32029844 on Build GRCh38
rs769798590 Chr.20: 32029862-32029862 on Build GRCh38
rs574357994 Chr.20: 32029762-32029762 on Build GRCh38
rs759257701 Chr.20: 32029856-32029856 on Build GRCh38
rs773259079 Chr.20: 32029780-32029780 on Build GRCh38
rs773065531 Chr.20: 32029693-32029693 on Build GRCh38
rs566762759 Chr.20: 32029704-32029704 on Build GRCh38
rs757803647 Chr.20: 32029816-32029816 on Build GRCh38
rs982100343 Chr.20: 32029679-32029679 on Build GRCh38
rs749780270 Chr.20: 32029752-32029752 on Build GRCh38
rs765274834 Chr.20: 32029714-32029714 on Build GRCh38
rs777221647 Chr.20: 32029744-32029744 on Build GRCh38
rs752827942 Chr.20: 32029722-32029722 on Build GRCh38
rs1051263534 Chr.20: 32029859-32029859 on Build GRCh38
rs754525891 Chr.20: 32029708-32029708 on Build GRCh38
rs755548732 Chr.20: 32029821-32029821 on Build GRCh38
rs776619849 Chr.20: 32029804-32029804 on Build GRCh38
rs557036107 Chr.20: 32029684-32029684 on Build GRCh38
rs140200202 Chr.20: 32029748-32029748 on Build GRCh38
rs371464754 Chr.20: 32029703-32029703 on Build GRCh38
rs868185994 Chr.20: 32029723-32029723 on Build GRCh38
rs774474624 Chr.20: 32029761-32029761 on Build GRCh38
rs780559369 Chr.20: 32029818-32029818 on Build GRCh38
rs78544950 Chr.20: 32029817-32029817 on Build GRCh38
rs747231679 Chr.20: 32029848-32029848 on Build GRCh38
rs552771372 Chr.20: 32029760-32029760 on Build GRCh38
rs765023826 Chr.20: 32029811-32029811 on Build GRCh38
rs779250286 Chr.20: 32029834-32029834 on Build GRCh38
rs751630594 Chr.20: 32029847-32029847 on Build GRCh38

View Full Product Details