Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 197421164-197421389 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRB1
Entrez Gene ID
23418
Gene Name
crumbs 1, cell polarity complex component
Gene Aliases
LCA8, RP12
UniGene
Hs.126135
Gene Chromosome Location
Chr.1: 197201462-197478455 on Build GRCh38
Transcripts
NM_001193640.1 NM_001257965.1 NM_001257966.1 NM_201253.2 NR_047563.1 NR_047564.1 XM_011509365.2 XM_011509367.1 XM_011509369.2 XM_017000851.1 XM_017000852.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6123821 c.1337A>T p.Q446L Chr.1: 197421372-197421372 on Build GRCh38
3481062 c.1397G>A p.G466E Chr.1: 197421225-197421225 on Build GRCh38
5379064 c.1273C>T p.L425L Chr.1: 197421308-197421308 on Build GRCh38
6534138 c.1342C>G p.Q448E Chr.1: 197421170-197421170 on Build GRCh38
901758 c.1422C>T p.G474G Chr.1: 197421250-197421250 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs898324034 Chr.1: 197421282-197421282 on Build GRCh38
rs963201816 Chr.1: 197421264-197421264 on Build GRCh38
rs200202087 Chr.1: 197421283-197421283 on Build GRCh38
rs142224492 Chr.1: 197421361-197421361 on Build GRCh38
rs150894028 Chr.1: 197421274-197421274 on Build GRCh38
rs776106605 Chr.1: 197421214-197421214 on Build GRCh38
rs761872993 Chr.1: 197421259-197421259 on Build GRCh38
rs769385495 Chr.1: 197421341-197421341 on Build GRCh38
rs985484481 Chr.1: 197421217-197421217 on Build GRCh38
rs917528638 Chr.1: 197421348-197421348 on Build GRCh38
rs776217552 Chr.1: 197421197-197421197 on Build GRCh38
rs866875342 Chr.1: 197421200-197421200 on Build GRCh38
rs375590765 Chr.1: 197421335-197421335 on Build GRCh38
rs863223342 Chr.1: 197421287-197421288 on Build GRCh38
rs937362854 Chr.1: 197421299-197421299 on Build GRCh38
rs770919155 Chr.1: 197421307-197421307 on Build GRCh38
rs763543345 Chr.1: 197421220-197421220 on Build GRCh38
rs759101736 Chr.1: 197421203-197421203 on Build GRCh38
rs146120995 Chr.1: 197421275-197421275 on Build GRCh38
rs777377174 Chr.1: 197421291-197421291 on Build GRCh38
rs767638739 Chr.1: 197421286-197421286 on Build GRCh38
rs62636282 Chr.1: 197421256-197421256 on Build GRCh38
rs767721094 Chr.1: 197421261-197421261 on Build GRCh38
rs372844443 Chr.1: 197421280-197421280 on Build GRCh38
rs745552217 Chr.1: 197421331-197421331 on Build GRCh38
rs3902057 Chr.1: 197421238-197421238 on Build GRCh38
rs866822473 Chr.1: 197421257-197421257 on Build GRCh38
rs62636265 Chr.1: 197421267-197421267 on Build GRCh38
rs780580887 Chr.1: 197421272-197421280 on Build GRCh38
rs62636264 Chr.1: 197421266-197421266 on Build GRCh38
rs144029476 Chr.1: 197421278-197421278 on Build GRCh38
rs769475443 Chr.1: 197421213-197421213 on Build GRCh38
rs750526087 Chr.1: 197421301-197421301 on Build GRCh38
rs758387370 Chr.1: 197421284-197421284 on Build GRCh38
rs35193230 Chr.1: 197421298-197421298 on Build GRCh38
rs992770731 Chr.1: 197421321-197421321 on Build GRCh38
rs530821170 Chr.1: 197421308-197421308 on Build GRCh38

View Full Product Details