Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 23048148-23048397 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDH24
Entrez Gene ID
64403
Gene Name
cadherin 24
Gene Aliases
CDH11L
UniGene
Hs.155912
Gene Chromosome Location
Chr.14: 23047061-23057619 on Build GRCh38
Transcripts
NM_022478.3 NM_144985.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6140254 c.2149C>T p.P717S Chr.14: 23048291-23048291 on Build GRCh38
3386474 c.2247C>T p.R749R Chr.14: 23048193-23048193 on Build GRCh38
6052581 c.2072C>T p.S691L Chr.14: 23048254-23048254 on Build GRCh38
1607507 c.2240G>A p.R747Q Chr.14: 23048200-23048200 on Build GRCh38
6625009 c.2164G>A p.V722M Chr.14: 23048276-23048276 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773253323 Chr.14: 23048284-23048284 on Build GRCh38
rs140801192 Chr.14: 23048325-23048325 on Build GRCh38
rs778843471 Chr.14: 23048275-23048275 on Build GRCh38
rs372007178 Chr.14: 23048166-23048166 on Build GRCh38
rs868226462 Chr.14: 23048186-23048186 on Build GRCh38
rs749727483 Chr.14: 23048270-23048270 on Build GRCh38
rs759004013 Chr.14: 23048337-23048337 on Build GRCh38
rs760230215 Chr.14: 23048351-23048351 on Build GRCh38
rs199930802 Chr.14: 23048177-23048177 on Build GRCh38
rs769693138 Chr.14: 23048313-23048313 on Build GRCh38
rs757640126 Chr.14: 23048309-23048309 on Build GRCh38
rs767105892 Chr.14: 23048298-23048298 on Build GRCh38
rs765787521 Chr.14: 23048218-23048218 on Build GRCh38
rs754617805 Chr.14: 23048250-23048250 on Build GRCh38
rs749936909 Chr.14: 23048178-23048178 on Build GRCh38
rs778302162 Chr.14: 23048266-23048266 on Build GRCh38
rs761662668 Chr.14: 23048281-23048281 on Build GRCh38
rs751286257 Chr.14: 23048241-23048241 on Build GRCh38
rs556244988 Chr.14: 23048176-23048176 on Build GRCh38
rs775170550 Chr.14: 23048339-23048339 on Build GRCh38
rs536011849 Chr.14: 23048285-23048285 on Build GRCh38
rs765984044 Chr.14: 23048293-23048293 on Build GRCh38
rs773871485 Chr.14: 23048334-23048334 on Build GRCh38
rs987348404 Chr.14: 23048214-23048214 on Build GRCh38
rs754425708 Chr.14: 23048306-23048306 on Build GRCh38
rs975462586 Chr.14: 23048228-23048228 on Build GRCh38
rs756632244 Chr.14: 23048265-23048265 on Build GRCh38
rs921267237 Chr.14: 23048231-23048231 on Build GRCh38
rs911692283 Chr.14: 23048215-23048215 on Build GRCh38
rs766981461 Chr.14: 23048338-23048338 on Build GRCh38
rs375215742 Chr.14: 23048354-23048354 on Build GRCh38
rs745895687 Chr.14: 23048310-23048310 on Build GRCh38
rs201603417 Chr.14: 23048355-23048355 on Build GRCh38
rs1054186520 Chr.14: 23048235-23048235 on Build GRCh38
rs539548365 Chr.14: 23048175-23048175 on Build GRCh38
rs150958098 Chr.14: 23048314-23048314 on Build GRCh38
rs781539258 Chr.14: 23048258-23048258 on Build GRCh38
rs534118854 Chr.14: 23048305-23048305 on Build GRCh38
rs766892810 Chr.14: 23048305-23048305 on Build GRCh38
rs867269217 Chr.14: 23048188-23048188 on Build GRCh38
rs752398646 Chr.14: 23048304-23048305 on Build GRCh38
rs756509445 Chr.14: 23048304-23048304 on Build GRCh38
rs759198282 Chr.14: 23048294-23048294 on Build GRCh38
rs936693122 Chr.14: 23048232-23048232 on Build GRCh38
rs771090320 Chr.14: 23048273-23048273 on Build GRCh38
rs776504296 Chr.14: 23048174-23048174 on Build GRCh38
rs762568844 Chr.14: 23048217-23048217 on Build GRCh38
rs746093683 Chr.14: 23048277-23048277 on Build GRCh38
rs776592627 Chr.14: 23048278-23048278 on Build GRCh38
rs752997456 Chr.14: 23048264-23048264 on Build GRCh38
rs867775788 Chr.14: 23048167-23048167 on Build GRCh38
rs780467111 Chr.14: 23048311-23048311 on Build GRCh38
rs754094790 Chr.14: 23048363-23048363 on Build GRCh38
rs755734438 Chr.14: 23048355-23048355 on Build GRCh38
rs759121232 Chr.14: 23048293-23048308 on Build GRCh38
rs914185680 Chr.14: 23048247-23048247 on Build GRCh38

View Full Product Details