Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 70833686-70833947 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HYDIN
Entrez Gene ID
54768
Gene Name
HYDIN, axonemal central pair apparatus protein
Gene Aliases
CILD5, HYDIN1, HYDIN2, PPP1R31
UniGene
Hs.351914
Gene Chromosome Location
Chr.16: 70802084-71230722 on Build GRCh38
Transcripts
NM_001270974.2 XM_006721206.2 XM_011523146.2 XM_011523147.1 XM_011523148.1 XM_011523151.1 XM_011523152.1 XM_011523155.2 XM_017023346.1 XM_017023347.1 XM_017023348.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1379347 c.13518C>T p.G4506G Chr.16: 70833901-70833901 on Build GRCh38
4411734 c.13517G>C p.G4506A Chr.16: 70833902-70833902 on Build GRCh38
4465049 c.13503C>T p.L4501L Chr.16: 70833916-70833916 on Build GRCh38
6892080 c.13511A>G p.N4504S Chr.16: 70833908-70833908 on Build GRCh38
6320530 c.13522G>T p.V4508L Chr.16: 70833897-70833897 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766770665 Chr.16: 70833874-70833874 on Build GRCh38
rs777444703 Chr.16: 70833900-70833900 on Build GRCh38
rs369579945 Chr.16: 70833877-70833877 on Build GRCh38
rs60408763 Chr.16: 70833768-70833768 on Build GRCh38
rs770831302 Chr.16: 70833916-70833916 on Build GRCh38
rs776629571 Chr.16: 70833920-70833920 on Build GRCh38
rs200226975 Chr.16: 70833876-70833876 on Build GRCh38
rs567495446 Chr.16: 70833832-70833832 on Build GRCh38
rs767722408 Chr.16: 70833851-70833851 on Build GRCh38
rs747626050 Chr.16: 70833880-70833880 on Build GRCh38
rs1049116002 Chr.16: 70833803-70833803 on Build GRCh38
rs949440087 Chr.16: 70833751-70833751 on Build GRCh38
rs372122774 Chr.16: 70833881-70833881 on Build GRCh38
rs887315035 Chr.16: 70833813-70833814 on Build GRCh38
rs1770442 Chr.16: 70833912-70833912 on Build GRCh38
rs965798498 Chr.16: 70833867-70833867 on Build GRCh38
rs995783152 Chr.16: 70833767-70833767 on Build GRCh38
rs750631110 Chr.16: 70833868-70833868 on Build GRCh38
rs756459434 Chr.16: 70833872-70833872 on Build GRCh38
rs201203817 Chr.16: 70833924-70833924 on Build GRCh38
rs374868182 Chr.16: 70833915-70833915 on Build GRCh38
rs535818619 Chr.16: 70833845-70833845 on Build GRCh38
rs1798372 Chr.16: 70833928-70833928 on Build GRCh38
rs1004368257 Chr.16: 70833814-70833814 on Build GRCh38
rs905151426 Chr.16: 70833766-70833766 on Build GRCh38
rs769787964 Chr.16: 70833926-70833926 on Build GRCh38
rs778386833 Chr.16: 70833878-70833878 on Build GRCh38
rs917985180 Chr.16: 70833720-70833720 on Build GRCh38

View Full Product Details