Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 89056901-89057172 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GBP1
Entrez Gene ID
2633
Gene Name
guanylate binding protein 1
Gene Aliases
-
UniGene
Hs.62661
Gene Chromosome Location
Chr.1: 89052304-89065360 on Build GRCh38
Transcripts
NM_002053.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5847443 c.889G>A p.V297M Chr.1: 89057120-89057120 on Build GRCh38
1344744 c.1046C>G p.T349S Chr.1: 89056963-89056963 on Build GRCh38
7117489 c.877C>G p.L293V Chr.1: 89057132-89057132 on Build GRCh38
912956 c.927G>A p.L309L Chr.1: 89057082-89057082 on Build GRCh38
6584082 c.1081G>A p.E361K Chr.1: 89056928-89056928 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373189716 Chr.1: 89057120-89057120 on Build GRCh38
rs780123997 Chr.1: 89057123-89057123 on Build GRCh38
rs774585837 Chr.1: 89056957-89056957 on Build GRCh38
rs777921995 Chr.1: 89057138-89057138 on Build GRCh38
rs17487439 Chr.1: 89057079-89057079 on Build GRCh38
rs769875691 Chr.1: 89057057-89057057 on Build GRCh38
rs745742295 Chr.1: 89057056-89057056 on Build GRCh38
rs763729522 Chr.1: 89057106-89057106 on Build GRCh38
rs745870988 Chr.1: 89056954-89056954 on Build GRCh38
rs756951201 Chr.1: 89057109-89057109 on Build GRCh38
rs1048425 Chr.1: 89056963-89056963 on Build GRCh38
rs757001279 Chr.1: 89057032-89057032 on Build GRCh38
rs771228081 Chr.1: 89057091-89057091 on Build GRCh38
rs144332469 Chr.1: 89057110-89057110 on Build GRCh38
rs779097744 Chr.1: 89057066-89057066 on Build GRCh38
rs746746558 Chr.1: 89056944-89056944 on Build GRCh38
rs759991663 Chr.1: 89057095-89057095 on Build GRCh38
rs781264867 Chr.1: 89056953-89056953 on Build GRCh38
rs748148281 Chr.1: 89057067-89057067 on Build GRCh38
rs778247235 Chr.1: 89057070-89057070 on Build GRCh38
rs759275064 Chr.1: 89056939-89056940 on Build GRCh38
rs749313065 Chr.1: 89057127-89057127 on Build GRCh38
rs764674329 Chr.1: 89057008-89057008 on Build GRCh38
rs768793043 Chr.1: 89056955-89056955 on Build GRCh38
rs150543608 Chr.1: 89057099-89057099 on Build GRCh38
rs745367467 Chr.1: 89057101-89057101 on Build GRCh38
rs561136888 Chr.1: 89057133-89057133 on Build GRCh38
rs757963839 Chr.1: 89057015-89057015 on Build GRCh38
rs926385480 Chr.1: 89057122-89057122 on Build GRCh38
rs777581694 Chr.1: 89056943-89056943 on Build GRCh38
rs377037156 Chr.1: 89057001-89057001 on Build GRCh38
rs770812253 Chr.1: 89056947-89056947 on Build GRCh38
rs545273025 Chr.1: 89057031-89057031 on Build GRCh38
rs781143681 Chr.1: 89057043-89057043 on Build GRCh38
rs778545272 Chr.1: 89057082-89057082 on Build GRCh38
rs149073593 Chr.1: 89056936-89056936 on Build GRCh38
rs766833210 Chr.1: 89057000-89057000 on Build GRCh38
rs577835962 Chr.1: 89057010-89057010 on Build GRCh38
rs150474721 Chr.1: 89057108-89057108 on Build GRCh38
rs1019603153 Chr.1: 89057075-89057075 on Build GRCh38
rs369692456 Chr.1: 89057105-89057105 on Build GRCh38
rs772216327 Chr.1: 89056967-89056967 on Build GRCh38
rs137876978 Chr.1: 89056992-89056992 on Build GRCh38
rs1028339586 Chr.1: 89056983-89056983 on Build GRCh38
rs140288607 Chr.1: 89056978-89056978 on Build GRCh38

View Full Product Details