Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 2027358-2027621 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GABRD
Entrez Gene ID
2563
Gene Name
gamma-aminobutyric acid type A receptor delta subunit
Gene Aliases
EIG10, EJM7, GEFSP5
UniGene
Hs.113882
Gene Chromosome Location
Chr.1: 2019329-2030753 on Build GRCh38
Transcripts
NM_000815.4 XM_011541194.2 XM_017000936.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1686884 c.493G>A p.D165N Chr.1: 2027599-2027599 on Build GRCh38
237430 c.492C>T p.C164C Chr.1: 2027598-2027598 on Build GRCh38
7369071 c.474C>G p.I158M Chr.1: 2027580-2027580 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1028800258 Chr.1: 2027401-2027401 on Build GRCh38
rs536807033 Chr.1: 2027404-2027404 on Build GRCh38
rs558900384 Chr.1: 2027521-2027521 on Build GRCh38
rs1018763758 Chr.1: 2027543-2027543 on Build GRCh38
rs113143432 Chr.1: 2027438-2027438 on Build GRCh38
rs567375126 Chr.1: 2027384-2027384 on Build GRCh38
rs752114528 Chr.1: 2027568-2027568 on Build GRCh38
rs989103106 Chr.1: 2027448-2027448 on Build GRCh38
rs987345076 Chr.1: 2027408-2027408 on Build GRCh38
rs375183388 Chr.1: 2027525-2027525 on Build GRCh38
rs539385265 Chr.1: 2027460-2027460 on Build GRCh38
rs28753776 Chr.1: 2027588-2027588 on Build GRCh38
rs367776284 Chr.1: 2027598-2027598 on Build GRCh38
rs954717883 Chr.1: 2027405-2027405 on Build GRCh38
rs996908287 Chr.1: 2027572-2027572 on Build GRCh38
rs937598383 Chr.1: 2027416-2027416 on Build GRCh38
rs762410432 Chr.1: 2027559-2027559 on Build GRCh38
rs528792200 Chr.1: 2027491-2027491 on Build GRCh38
rs150102040 Chr.1: 2027554-2027554 on Build GRCh38
rs750763018 Chr.1: 2027586-2027586 on Build GRCh38
rs899845390 Chr.1: 2027567-2027567 on Build GRCh38
rs189301522 Chr.1: 2027409-2027409 on Build GRCh38
rs955512295 Chr.1: 2027582-2027582 on Build GRCh38
rs763571219 Chr.1: 2027562-2027562 on Build GRCh38
rs767993168 Chr.1: 2027574-2027574 on Build GRCh38
rs1044806955 Chr.1: 2027473-2027473 on Build GRCh38
rs887351818 Chr.1: 2027533-2027533 on Build GRCh38
rs896011418 Chr.1: 2027440-2027440 on Build GRCh38
rs1043098354 Chr.1: 2027387-2027387 on Build GRCh38
rs947772932 Chr.1: 2027464-2027464 on Build GRCh38
rs752352993 Chr.1: 2027556-2027556 on Build GRCh38
rs756455348 Chr.1: 2027589-2027589 on Build GRCh38
rs987988338 Chr.1: 2027557-2027557 on Build GRCh38
rs1027734760 Chr.1: 2027509-2027509 on Build GRCh38
rs756144316 Chr.1: 2027498-2027498 on Build GRCh38
rs776563999 Chr.1: 2027526-2027526 on Build GRCh38
rs748813137 Chr.1: 2027569-2027569 on Build GRCh38
rs181895553 Chr.1: 2027550-2027550 on Build GRCh38
rs28437090 Chr.1: 2027583-2027583 on Build GRCh38
rs923840254 Chr.1: 2027420-2027420 on Build GRCh38
rs914911532 Chr.1: 2027449-2027449 on Build GRCh38

View Full Product Details