Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 139362450-139362705 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COPB2
Entrez Gene ID
9276
Gene Name
coatomer protein complex subunit beta 2
Gene Aliases
beta'-COP
UniGene
Hs.681616
Gene Chromosome Location
Chr.3: 139355595-139389680 on Build GRCh38
Transcripts
NM_004766.2 NR_023350.1 XM_017007512.1 XM_017007513.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4444887 c.1938G>T p.E646D Chr.3: 139362464-139362464 on Build GRCh38
3124284 c.1913C>T p.S638F Chr.3: 139362489-139362489 on Build GRCh38
6281778 c.1939C>A p.L647I Chr.3: 139362463-139362463 on Build GRCh38
5918036 c.1930C>T p.R644C Chr.3: 139362472-139362472 on Build GRCh38
6468489 c.1909G>A p.V637I Chr.3: 139362493-139362493 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776424276 Chr.3: 139362575-139362578 on Build GRCh38
rs34944211 Chr.3: 139362651-139362652 on Build GRCh38
rs796471002 Chr.3: 139362589-139362590 on Build GRCh38
rs747326058 Chr.3: 139362554-139362554 on Build GRCh38
rs767436146 Chr.3: 139362556-139362556 on Build GRCh38
rs764014600 Chr.3: 139362494-139362494 on Build GRCh38
rs764099279 Chr.3: 139362471-139362471 on Build GRCh38
rs572765652 Chr.3: 139362538-139362541 on Build GRCh38
rs528065676 Chr.3: 139362493-139362493 on Build GRCh38
rs779026005 Chr.3: 139362532-139362532 on Build GRCh38
rs780219618 Chr.3: 139362544-139362544 on Build GRCh38
rs1053261040 Chr.3: 139362571-139362571 on Build GRCh38
rs561382494 Chr.3: 139362673-139362673 on Build GRCh38
rs752359547 Chr.3: 139362490-139362490 on Build GRCh38
rs759073703 Chr.3: 139362482-139362482 on Build GRCh38
rs947082655 Chr.3: 139362525-139362525 on Build GRCh38
rs149520242 Chr.3: 139362506-139362506 on Build GRCh38
rs113303328 Chr.3: 139362517-139362517 on Build GRCh38
rs745473877 Chr.3: 139362577-139362578 on Build GRCh38
rs905369799 Chr.3: 139362590-139362591 on Build GRCh38
rs750382905 Chr.3: 139362563-139362563 on Build GRCh38
rs889559862 Chr.3: 139362562-139362562 on Build GRCh38
rs138037779 Chr.3: 139362495-139362495 on Build GRCh38
rs759418803 Chr.3: 139362537-139362538 on Build GRCh38
rs773888231 Chr.3: 139362472-139362472 on Build GRCh38
rs750338124 Chr.3: 139362540-139362540 on Build GRCh38
rs377700267 Chr.3: 139362551-139362551 on Build GRCh38
rs182787422 Chr.3: 139362543-139362543 on Build GRCh38
rs187760995 Chr.3: 139362589-139362589 on Build GRCh38
rs142305414 Chr.3: 139362486-139362486 on Build GRCh38
rs897032206 Chr.3: 139362652-139362652 on Build GRCh38
rs57549985 Chr.3: 139362575-139362576 on Build GRCh38
rs752446301 Chr.3: 139362562-139362562 on Build GRCh38
rs893380414 Chr.3: 139362481-139362481 on Build GRCh38
rs892926453 Chr.3: 139362574-139362575 on Build GRCh38
rs63118562 Chr.3: 139362586-139362587 on Build GRCh38
rs1019449466 Chr.3: 139362601-139362601 on Build GRCh38
rs1022386212 Chr.3: 139362589-139362590 on Build GRCh38
rs1011021241 Chr.3: 139362584-139362586 on Build GRCh38

View Full Product Details