Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 50441337-50441610 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAF1
Entrez Gene ID
11124
Gene Name
Fas associated factor 1
Gene Aliases
CGI-03, HFAF1s, UBXD12, UBXN3A, hFAF1
UniGene
Hs.530402
Gene Chromosome Location
Chr.1: 50441263-50960264 on Build GRCh38
Transcripts
NM_007051.2 XM_017000136.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5769802 c.1919C>A p.P640H Chr.1: 50441474-50441474 on Build GRCh38
3805501 c.1876C>T p.Q626* Chr.1: 50441517-50441517 on Build GRCh38
5232417 c.1150C>T p.Q384* Chr.1: 50441517-50441517 on Build GRCh38
3805500 c.1948G>C p.E650Q Chr.1: 50441445-50441445 on Build GRCh38
5203377 c.1222G>C p.E408Q Chr.1: 50441445-50441445 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767077547 Chr.1: 50441536-50441536 on Build GRCh38
rs911470911 Chr.1: 50441388-50441388 on Build GRCh38
rs1048718172 Chr.1: 50441381-50441381 on Build GRCh38
rs772212793 Chr.1: 50441405-50441405 on Build GRCh38
rs749918289 Chr.1: 50441574-50441574 on Build GRCh38
rs764699238 Chr.1: 50441389-50441389 on Build GRCh38
rs747294487 Chr.1: 50441435-50441435 on Build GRCh38
rs759777870 Chr.1: 50441438-50441438 on Build GRCh38
rs775917503 Chr.1: 50441482-50441482 on Build GRCh38
rs543370954 Chr.1: 50441423-50441423 on Build GRCh38
rs559818433 Chr.1: 50441386-50441386 on Build GRCh38
rs761313785 Chr.1: 50441515-50441515 on Build GRCh38
rs533521926 Chr.1: 50441426-50441426 on Build GRCh38
rs1041201784 Chr.1: 50441408-50441408 on Build GRCh38
rs763528596 Chr.1: 50441371-50441371 on Build GRCh38
rs765795263 Chr.1: 50441439-50441439 on Build GRCh38
rs143311114 Chr.1: 50441564-50441564 on Build GRCh38
rs191382480 Chr.1: 50441370-50441370 on Build GRCh38
rs200710823 Chr.1: 50441436-50441436 on Build GRCh38
rs563037274 Chr.1: 50441569-50441569 on Build GRCh38
rs773422449 Chr.1: 50441407-50441407 on Build GRCh38

View Full Product Details