Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 54360482-54360723 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C18orf54
Entrez Gene ID
162681
Gene Name
chromosome 18 open reading frame 54
Gene Aliases
LAS2
UniGene
Hs.208701
Gene Chromosome Location
Chr.18: 54357914-54382035 on Build GRCh38
Transcripts
NM_001288980.1 NM_001288981.1 NM_001288982.1 NM_173529.5 NR_110234.1 XM_005258201.1 XM_005258203.1 XM_017025567.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5799147 c.52T>G p.S18A Chr.18: 54360624-54360624 on Build GRCh38
7220403 c.83G>A p.G28D Chr.18: 54360655-54360655 on Build GRCh38
7218041 c.109T>C p.S37P Chr.18: 54360681-54360681 on Build GRCh38
988979 c.6G>A p.A2A Chr.18: 54360578-54360578 on Build GRCh38
4072593 c.148C>T p.Q50* Chr.18: 54360720-54360720 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761468606 Chr.18: 54360678-54360678 on Build GRCh38
rs376517433 Chr.18: 54360552-54360552 on Build GRCh38
rs145235987 Chr.18: 54360630-54360630 on Build GRCh38
rs150551052 Chr.18: 54360635-54360635 on Build GRCh38
rs139710634 Chr.18: 54360578-54360578 on Build GRCh38
rs967258081 Chr.18: 54360652-54360652 on Build GRCh38
rs114831324 Chr.18: 54360683-54360683 on Build GRCh38
rs756713933 Chr.18: 54360596-54360596 on Build GRCh38
rs367885391 Chr.18: 54360565-54360565 on Build GRCh38
rs866073111 Chr.18: 54360690-54360690 on Build GRCh38
rs746294693 Chr.18: 54360640-54360640 on Build GRCh38
rs766385586 Chr.18: 54360689-54360689 on Build GRCh38
rs780701874 Chr.18: 54360598-54360598 on Build GRCh38
rs773943063 Chr.18: 54360677-54360677 on Build GRCh38
rs770399456 Chr.18: 54360644-54360644 on Build GRCh38
rs776868526 Chr.18: 54360636-54360636 on Build GRCh38
rs749164143 Chr.18: 54360620-54360620 on Build GRCh38
rs748904824 Chr.18: 54360554-54360554 on Build GRCh38
rs763426642 Chr.18: 54360667-54360667 on Build GRCh38
rs780156014 Chr.18: 54360600-54360600 on Build GRCh38
rs371866457 Chr.18: 54360542-54360542 on Build GRCh38
rs756079510 Chr.18: 54360601-54360601 on Build GRCh38
rs779705365 Chr.18: 54360604-54360604 on Build GRCh38
rs369850714 Chr.18: 54360526-54360528 on Build GRCh38
rs533744271 Chr.18: 54360632-54360632 on Build GRCh38
rs753548344 Chr.18: 54360692-54360692 on Build GRCh38
rs760114584 Chr.18: 54360573-54360573 on Build GRCh38
rs760889743 Chr.18: 54360559-54360559 on Build GRCh38
rs910144743 Chr.18: 54360669-54360669 on Build GRCh38
rs3819129 Chr.18: 54360655-54360655 on Build GRCh38
rs766946669 Chr.18: 54360681-54360681 on Build GRCh38
rs763795358 Chr.18: 54360595-54360595 on Build GRCh38
rs118019249 Chr.18: 54360512-54360512 on Build GRCh38
rs201779097 Chr.18: 54360672-54360672 on Build GRCh38
rs200997925 Chr.18: 54360555-54360555 on Build GRCh38
rs771536342 Chr.18: 54360562-54360562 on Build GRCh38
rs758669294 Chr.18: 54360628-54360628 on Build GRCh38
rs145211389 Chr.18: 54360695-54360695 on Build GRCh38
rs536394320 Chr.18: 54360577-54360577 on Build GRCh38
rs77952069 Chr.18: 54360682-54360682 on Build GRCh38
rs762510584 Chr.18: 54360589-54360589 on Build GRCh38

View Full Product Details