Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 10419721-10419946 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HPCAL1
Entrez Gene ID
3241
Gene Name
hippocalcin like 1
Gene Aliases
BDR1, HLP2, VILIP-3
UniGene
Hs.580427
Gene Chromosome Location
Chr.2: 10302124-10427617 on Build GRCh38
Transcripts
NM_001258357.1 NM_001258358.1 NM_001258359.1 NM_002149.3 NM_134421.2 XM_005246160.1 XM_005246161.1 XM_005246162.1 XM_005246163.1 XM_011510346.2 XM_011510347.1 XM_011510348.1 XM_011510349.1 XM_017003950.1 XM_017003951.1 XM_017003952.1 XM_017003953.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4083923 c.150G>A p.K50K Chr.2: 10419907-10419907 on Build GRCh38
1004736 c.95A>G p.K32R Chr.2: 10419852-10419852 on Build GRCh38
440927 c.73C>T p.H25Y Chr.2: 10419830-10419830 on Build GRCh38
1004737 c.157G>A p.A53T Chr.2: 10419914-10419914 on Build GRCh38
1209928 c.16A>C p.S6R Chr.2: 10419773-10419773 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749574883 Chr.2: 10419902-10419904 on Build GRCh38
rs746105701 Chr.2: 10419793-10419793 on Build GRCh38
rs766882950 Chr.2: 10419884-10419884 on Build GRCh38
rs969150850 Chr.2: 10419791-10419791 on Build GRCh38
rs761644160 Chr.2: 10419826-10419826 on Build GRCh38
rs762644018 Chr.2: 10419865-10419865 on Build GRCh38
rs143370055 Chr.2: 10419877-10419877 on Build GRCh38
rs1045519641 Chr.2: 10419859-10419859 on Build GRCh38
rs756557225 Chr.2: 10419768-10419768 on Build GRCh38
rs1023291597 Chr.2: 10419749-10419749 on Build GRCh38
rs778319187 Chr.2: 10419895-10419895 on Build GRCh38
rs530325936 Chr.2: 10419889-10419889 on Build GRCh38
rs751061337 Chr.2: 10419742-10419742 on Build GRCh38
rs3755260 Chr.2: 10419878-10419878 on Build GRCh38
rs375884714 Chr.2: 10419914-10419914 on Build GRCh38
rs769646547 Chr.2: 10419830-10419830 on Build GRCh38
rs532435148 Chr.2: 10419784-10419784 on Build GRCh38
rs550682647 Chr.2: 10419825-10419825 on Build GRCh38
rs771244006 Chr.2: 10419808-10419808 on Build GRCh38
rs757645800 Chr.2: 10419919-10419919 on Build GRCh38
rs749584785 Chr.2: 10419807-10419807 on Build GRCh38
rs778908511 Chr.2: 10419817-10419817 on Build GRCh38
rs376171717 Chr.2: 10419741-10419741 on Build GRCh38
rs565024473 Chr.2: 10419754-10419754 on Build GRCh38
rs915152688 Chr.2: 10419903-10419903 on Build GRCh38
rs772961399 Chr.2: 10419853-10419853 on Build GRCh38
rs200622229 Chr.2: 10419744-10419744 on Build GRCh38
rs778267644 Chr.2: 10419782-10419782 on Build GRCh38
rs759094665 Chr.2: 10419883-10419883 on Build GRCh38
rs1034740196 Chr.2: 10419827-10419827 on Build GRCh38
rs951719147 Chr.2: 10419925-10419925 on Build GRCh38
rs756679860 Chr.2: 10419893-10419893 on Build GRCh38
rs781739211 Chr.2: 10419745-10419745 on Build GRCh38

View Full Product Details