Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 26188850-26189115 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HIST1H4D
Entrez Gene ID
8360
Gene Name
histone cluster 1, H4d
Gene Aliases
H4/b, H4FB, dJ221C16.9
UniGene
Hs.248179
Gene Chromosome Location
Chr.6: 26188710-26189076 on Build GRCh38
Transcripts
NM_003539.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6201272 c.194A>G p.N65S Chr.6: 26188883-26188883 on Build GRCh38
6173449 c.100G>A p.A34T Chr.6: 26188977-26188977 on Build GRCh38
4390940 c.177G>A p.L59L Chr.6: 26188900-26188900 on Build GRCh38
1254227 c.92C>T p.T31I Chr.6: 26188985-26188985 on Build GRCh38
6680345 c.63G>C p.K21N Chr.6: 26189014-26189014 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755124179 Chr.6: 26188890-26188890 on Build GRCh38
rs373964325 Chr.6: 26188944-26188944 on Build GRCh38
rs202049591 Chr.6: 26188983-26188983 on Build GRCh38
rs756481227 Chr.6: 26188957-26188957 on Build GRCh38
rs201318128 Chr.6: 26188918-26188918 on Build GRCh38
rs776014318 Chr.6: 26189012-26189012 on Build GRCh38
rs745766433 Chr.6: 26189013-26189013 on Build GRCh38
rs752756387 Chr.6: 26188894-26188894 on Build GRCh38
rs750906087 Chr.6: 26189035-26189046 on Build GRCh38
rs1019899529 Chr.6: 26189083-26189083 on Build GRCh38
rs369583517 Chr.6: 26188911-26188911 on Build GRCh38
rs544244183 Chr.6: 26188875-26188875 on Build GRCh38
rs773088974 Chr.6: 26188959-26188959 on Build GRCh38
rs375888188 Chr.6: 26189029-26189029 on Build GRCh38
rs1027364251 Chr.6: 26188882-26188883 on Build GRCh38
rs757687288 Chr.6: 26188948-26188948 on Build GRCh38
rs746577882 Chr.6: 26188972-26188972 on Build GRCh38
rs746297564 Chr.6: 26188910-26188910 on Build GRCh38
rs199624269 Chr.6: 26189068-26189068 on Build GRCh38
rs750321677 Chr.6: 26188880-26188880 on Build GRCh38
rs760676646 Chr.6: 26188885-26188885 on Build GRCh38
rs539823981 Chr.6: 26189063-26189063 on Build GRCh38
rs142303259 Chr.6: 26189050-26189050 on Build GRCh38
rs574527741 Chr.6: 26189058-26189058 on Build GRCh38
rs1004567395 Chr.6: 26189047-26189048 on Build GRCh38
rs386698221 Chr.6: 26189084-26189085 on Build GRCh38
rs780528340 Chr.6: 26188998-26188998 on Build GRCh38
rs113589835 Chr.6: 26189077-26189077 on Build GRCh38
rs761193902 Chr.6: 26189035-26189036 on Build GRCh38
rs149672595 Chr.6: 26188877-26188877 on Build GRCh38
rs146500115 Chr.6: 26189023-26189023 on Build GRCh38
rs752846007 Chr.6: 26188934-26188934 on Build GRCh38
rs766377500 Chr.6: 26188930-26188930 on Build GRCh38
rs201162303 Chr.6: 26188999-26188999 on Build GRCh38
rs774402895 Chr.6: 26189061-26189061 on Build GRCh38
rs139439104 Chr.6: 26188876-26188876 on Build GRCh38
rs772816360 Chr.6: 26188969-26188969 on Build GRCh38
rs149993334 Chr.6: 26189075-26189075 on Build GRCh38
rs1034362018 Chr.6: 26188888-26188888 on Build GRCh38
rs1002632495 Chr.6: 26188966-26188966 on Build GRCh38
rs756656965 Chr.6: 26189038-26189038 on Build GRCh38
rs753095005 Chr.6: 26189080-26189080 on Build GRCh38
rs770403660 Chr.6: 26189007-26189007 on Build GRCh38
rs754242447 Chr.6: 26189042-26189042 on Build GRCh38
rs751583245 Chr.6: 26188995-26188995 on Build GRCh38
rs548178202 Chr.6: 26188981-26188981 on Build GRCh38
rs777538918 Chr.6: 26189002-26189002 on Build GRCh38
rs760487699 Chr.6: 26188927-26188927 on Build GRCh38
rs750884941 Chr.6: 26188997-26188997 on Build GRCh38
rs749175349 Chr.6: 26189070-26189070 on Build GRCh38
rs570837491 Chr.6: 26189018-26189018 on Build GRCh38
rs554616183 Chr.6: 26189046-26189046 on Build GRCh38
rs1011647555 Chr.6: 26188974-26188974 on Build GRCh38
rs771503236 Chr.6: 26188909-26188909 on Build GRCh38
rs560226680 Chr.6: 26189065-26189065 on Build GRCh38
rs1022698942 Chr.6: 26189078-26189078 on Build GRCh38
rs769708257 Chr.6: 26189053-26189053 on Build GRCh38
rs79868973 Chr.6: 26189084-26189084 on Build GRCh38
rs139257264 Chr.6: 26189044-26189044 on Build GRCh38
rs374273581 Chr.6: 26189089-26189089 on Build GRCh38
rs1004834260 Chr.6: 26188892-26188892 on Build GRCh38
rs201894000 Chr.6: 26188979-26188979 on Build GRCh38
rs767855745 Chr.6: 26189033-26189053 on Build GRCh38
rs368566658 Chr.6: 26189048-26189048 on Build GRCh38
rs747940205 Chr.6: 26189074-26189074 on Build GRCh38
rs768846426 Chr.6: 26188995-26188996 on Build GRCh38
rs746496736 Chr.6: 26189006-26189006 on Build GRCh38
rs140430462 Chr.6: 26189067-26189067 on Build GRCh38
rs754922841 Chr.6: 26189087-26189087 on Build GRCh38
rs747755990 Chr.6: 26188964-26188964 on Build GRCh38
rs746214961 Chr.6: 26188954-26188954 on Build GRCh38
rs1036250121 Chr.6: 26189062-26189062 on Build GRCh38
rs747454273 Chr.6: 26188903-26188903 on Build GRCh38
rs767500097 Chr.6: 26188996-26188996 on Build GRCh38
rs761302691 Chr.6: 26188891-26188891 on Build GRCh38
rs979943130 Chr.6: 26188917-26188917 on Build GRCh38
rs144085716 Chr.6: 26188987-26188987 on Build GRCh38
rs762079534 Chr.6: 26189030-26189031 on Build GRCh38
rs765437973 Chr.6: 26188932-26188932 on Build GRCh38
rs763133612 Chr.6: 26188989-26188989 on Build GRCh38
rs762893570 Chr.6: 26188960-26188960 on Build GRCh38
rs1038731202 Chr.6: 26188873-26188873 on Build GRCh38
rs780709577 Chr.6: 26189049-26189049 on Build GRCh38
rs774215046 Chr.6: 26188912-26188912 on Build GRCh38
rs914560070 Chr.6: 26188907-26188907 on Build GRCh38
rs758099718 Chr.6: 26189001-26189001 on Build GRCh38
rs779146331 Chr.6: 26189045-26189045 on Build GRCh38
rs753696049 Chr.6: 26188889-26188889 on Build GRCh38
rs767039326 Chr.6: 26189041-26189041 on Build GRCh38
rs781753823 Chr.6: 26188951-26188951 on Build GRCh38
rs754328775 Chr.6: 26189066-26189066 on Build GRCh38
rs762799179 Chr.6: 26189020-26189020 on Build GRCh38
rs368348095 Chr.6: 26188902-26188902 on Build GRCh38
rs978290741 Chr.6: 26188986-26188986 on Build GRCh38
rs984553001 Chr.6: 26189047-26189047 on Build GRCh38
rs971194645 Chr.6: 26188913-26188913 on Build GRCh38
rs371511135 Chr.6: 26188942-26188942 on Build GRCh38
rs569549632 Chr.6: 26188973-26188973 on Build GRCh38
rs73736557 Chr.6: 26188978-26188978 on Build GRCh38
rs200821779 Chr.6: 26189076-26189076 on Build GRCh38
rs774755862 Chr.6: 26189022-26189045 on Build GRCh38
rs768397391 Chr.6: 26188869-26188869 on Build GRCh38
rs141140958 Chr.6: 26189079-26189079 on Build GRCh38
rs368091679 Chr.6: 26188991-26188991 on Build GRCh38
rs552478518 Chr.6: 26188965-26188965 on Build GRCh38
rs146262007 Chr.6: 26188963-26188963 on Build GRCh38
rs151274627 Chr.6: 26189056-26189056 on Build GRCh38
rs201613383 Chr.6: 26189072-26189072 on Build GRCh38
rs772871879 Chr.6: 26188921-26188921 on Build GRCh38
rs78735537 Chr.6: 26189085-26189085 on Build GRCh38
rs749934539 Chr.6: 26189069-26189069 on Build GRCh38
rs150838709 Chr.6: 26189032-26189032 on Build GRCh38
rs763205100 Chr.6: 26188900-26188902 on Build GRCh38
rs200441194 Chr.6: 26189014-26189014 on Build GRCh38
rs779259978 Chr.6: 26188911-26188912 on Build GRCh38
rs765008574 Chr.6: 26188920-26188920 on Build GRCh38
rs759432453 Chr.6: 26188931-26188931 on Build GRCh38
rs776694850 Chr.6: 26188977-26188977 on Build GRCh38
rs1033729066 Chr.6: 26188929-26188929 on Build GRCh38
rs758536923 Chr.6: 26188900-26188900 on Build GRCh38
rs1040156315 Chr.6: 26188925-26188925 on Build GRCh38

View Full Product Details