Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 26641168-26641409 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CIB4
Entrez Gene ID
130106
Gene Name
calcium and integrin binding family member 4
Gene Aliases
KIP4
UniGene
Hs.591579
Gene Chromosome Location
Chr.2: 26581202-26641798 on Build GRCh38
Transcripts
NM_001029881.2 XM_011532514.2 XM_017003331.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4672427 c.3G>A p.M1I Chr.2: 26641312-26641312 on Build GRCh38
4777214 c.46G>C p.E16Q Chr.2: 26641269-26641269 on Build GRCh38
4093406 c.47A>T p.E16V Chr.2: 26641268-26641268 on Build GRCh38
720696 c.36G>A p.E12E Chr.2: 26641279-26641279 on Build GRCh38
720697 c.46G>A p.E16K Chr.2: 26641269-26641269 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200491839 Chr.2: 26641245-26641245 on Build GRCh38
rs141660203 Chr.2: 26641283-26641283 on Build GRCh38
rs143344446 Chr.2: 26641294-26641294 on Build GRCh38
rs750933487 Chr.2: 26641215-26641215 on Build GRCh38
rs142383569 Chr.2: 26641285-26641285 on Build GRCh38
rs778685891 Chr.2: 26641256-26641256 on Build GRCh38
rs147692073 Chr.2: 26641258-26641258 on Build GRCh38
rs770951282 Chr.2: 26641253-26641253 on Build GRCh38
rs750375496 Chr.2: 26641250-26641250 on Build GRCh38
rs199788270 Chr.2: 26641349-26641349 on Build GRCh38
rs755693055 Chr.2: 26641247-26641247 on Build GRCh38
rs773395158 Chr.2: 26641305-26641305 on Build GRCh38
rs755263351 Chr.2: 26641242-26641242 on Build GRCh38
rs771802864 Chr.2: 26641262-26641262 on Build GRCh38
rs781017089 Chr.2: 26641244-26641244 on Build GRCh38
rs774674937 Chr.2: 26641344-26641344 on Build GRCh38
rs1006213709 Chr.2: 26641383-26641383 on Build GRCh38
rs779243945 Chr.2: 26641327-26641327 on Build GRCh38
rs781513145 Chr.2: 26641326-26641329 on Build GRCh38
rs763816864 Chr.2: 26641366-26641366 on Build GRCh38
rs777496280 Chr.2: 26641249-26641249 on Build GRCh38
rs752001912 Chr.2: 26641241-26641241 on Build GRCh38
rs562782951 Chr.2: 26641369-26641369 on Build GRCh38
rs753112198 Chr.2: 26641358-26641358 on Build GRCh38
rs755889564 Chr.2: 26641362-26641362 on Build GRCh38
rs372812526 Chr.2: 26641384-26641384 on Build GRCh38
rs763580281 Chr.2: 26641224-26641224 on Build GRCh38
rs766947640 Chr.2: 26641322-26641322 on Build GRCh38
rs201259271 Chr.2: 26641281-26641281 on Build GRCh38
rs1053750402 Chr.2: 26641197-26641197 on Build GRCh38
rs899093560 Chr.2: 26641189-26641189 on Build GRCh38
rs1047662412 Chr.2: 26641367-26641367 on Build GRCh38
rs767930760 Chr.2: 26641352-26641352 on Build GRCh38
rs765242198 Chr.2: 26641214-26641214 on Build GRCh38
rs373642726 Chr.2: 26641227-26641227 on Build GRCh38
rs762917297 Chr.2: 26641315-26641315 on Build GRCh38

View Full Product Details