Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 115276886-115277135 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC112
Entrez Gene ID
153733
Gene Name
coiled-coil domain containing 112
Gene Aliases
MBC1
UniGene
Hs.436121
Gene Chromosome Location
Chr.5: 115267188-115296761 on Build GRCh38
Transcripts
NM_001040440.2 NM_152549.2 XM_005271908.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3714968 c.421C>T p.Q141* Chr.5: 115276995-115276995 on Build GRCh38
2991848 c.451+1G>T p.? Chr.5: 115276964-115276964 on Build GRCh38
1619336 c.400A>G p.N134D Chr.5: 115277016-115277016 on Build GRCh38
6256717 c.376C>A p.Q126K Chr.5: 115277040-115277040 on Build GRCh38
397808 c.403G>T p.V135L Chr.5: 115277013-115277013 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774337503 Chr.5: 115276951-115276951 on Build GRCh38
rs781506425 Chr.5: 115276978-115276978 on Build GRCh38
rs769990914 Chr.5: 115277094-115277094 on Build GRCh38
rs757669947 Chr.5: 115276968-115276968 on Build GRCh38
rs758908318 Chr.5: 115276955-115276955 on Build GRCh38
rs755781819 Chr.5: 115277015-115277015 on Build GRCh38
rs988889935 Chr.5: 115277095-115277095 on Build GRCh38
rs200075094 Chr.5: 115276959-115276959 on Build GRCh38
rs748768635 Chr.5: 115277035-115277035 on Build GRCh38
rs371062258 Chr.5: 115276960-115276960 on Build GRCh38
rs765291392 Chr.5: 115276948-115276948 on Build GRCh38
rs759795907 Chr.5: 115276927-115276927 on Build GRCh38
rs370612378 Chr.5: 115277051-115277051 on Build GRCh38
rs759690339 Chr.5: 115277075-115277075 on Build GRCh38
rs762969732 Chr.5: 115277104-115277104 on Build GRCh38
rs779447491 Chr.5: 115277021-115277021 on Build GRCh38
rs938782354 Chr.5: 115277082-115277082 on Build GRCh38
rs896170434 Chr.5: 115277009-115277009 on Build GRCh38
rs768748024 Chr.5: 115277046-115277046 on Build GRCh38
rs183051493 Chr.5: 115277071-115277071 on Build GRCh38
rs773042902 Chr.5: 115277072-115277072 on Build GRCh38
rs146328652 Chr.5: 115277014-115277014 on Build GRCh38
rs969686564 Chr.5: 115277044-115277044 on Build GRCh38
rs377146034 Chr.5: 115276998-115276998 on Build GRCh38
rs775462235 Chr.5: 115277102-115277102 on Build GRCh38
rs372585190 Chr.5: 115276941-115276945 on Build GRCh38
rs374319734 Chr.5: 115277057-115277057 on Build GRCh38
rs752763986 Chr.5: 115276951-115276951 on Build GRCh38
rs761919643 Chr.5: 115277095-115277096 on Build GRCh38

View Full Product Details