Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 63558283-63558556 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HRASLS2
Entrez Gene ID
54979
Gene Name
HRAS like suppressor 2
Gene Aliases
PLA1/2-2
UniGene
Hs.272805
Gene Chromosome Location
Chr.11: 63552759-63565072 on Build GRCh38
Transcripts
NM_017878.1 XM_011545120.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5468491 c.387+3G>A p.? Chr.11: 63558389-63558389 on Build GRCh38
5492811 c.284G>A p.R95Q Chr.11: 63558495-63558495 on Build GRCh38
366081 c.307G>T p.E103* Chr.11: 63558472-63558472 on Build GRCh38
7401264 c.322C>T p.L108L Chr.11: 63558457-63558457 on Build GRCh38
5532410 c.320C>T p.S107L Chr.11: 63558459-63558459 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201966137 Chr.11: 63558391-63558391 on Build GRCh38
rs200152066 Chr.11: 63558453-63558453 on Build GRCh38
rs148548609 Chr.11: 63558503-63558503 on Build GRCh38
rs966639135 Chr.11: 63558315-63558315 on Build GRCh38
rs781640640 Chr.11: 63558442-63558442 on Build GRCh38
rs369662368 Chr.11: 63558525-63558525 on Build GRCh38
rs762859896 Chr.11: 63558509-63558509 on Build GRCh38
rs995599513 Chr.11: 63558468-63558468 on Build GRCh38
rs147385363 Chr.11: 63558424-63558424 on Build GRCh38
rs756449411 Chr.11: 63558459-63558459 on Build GRCh38
rs867727364 Chr.11: 63558405-63558405 on Build GRCh38
rs746226646 Chr.11: 63558372-63558372 on Build GRCh38
rs771611099 Chr.11: 63558473-63558473 on Build GRCh38
rs758457110 Chr.11: 63558344-63558344 on Build GRCh38
rs200068825 Chr.11: 63558367-63558367 on Build GRCh38
rs966598888 Chr.11: 63558422-63558422 on Build GRCh38
rs183132185 Chr.11: 63558375-63558375 on Build GRCh38
rs775361402 Chr.11: 63558421-63558430 on Build GRCh38
rs772611818 Chr.11: 63558389-63558389 on Build GRCh38
rs199657668 Chr.11: 63558522-63558522 on Build GRCh38
rs138690084 Chr.11: 63558403-63558403 on Build GRCh38
rs777548929 Chr.11: 63558486-63558486 on Build GRCh38
rs372878699 Chr.11: 63558496-63558496 on Build GRCh38
rs747479207 Chr.11: 63558354-63558354 on Build GRCh38
rs921447808 Chr.11: 63558352-63558352 on Build GRCh38
rs924925227 Chr.11: 63558327-63558327 on Build GRCh38
rs757770333 Chr.11: 63558366-63558366 on Build GRCh38
rs776664376 Chr.11: 63558495-63558495 on Build GRCh38
rs764214220 Chr.11: 63558510-63558510 on Build GRCh38
rs1025344728 Chr.11: 63558314-63558314 on Build GRCh38
rs371200365 Chr.11: 63558418-63558418 on Build GRCh38
rs746491233 Chr.11: 63558487-63558487 on Build GRCh38
rs550940863 Chr.11: 63558380-63558380 on Build GRCh38
rs142689067 Chr.11: 63558409-63558409 on Build GRCh38
rs571117336 Chr.11: 63558381-63558381 on Build GRCh38
rs777742791 Chr.11: 63558346-63558346 on Build GRCh38
rs764966202 Chr.11: 63558413-63558413 on Build GRCh38
rs890103172 Chr.11: 63558385-63558385 on Build GRCh38
rs771546037 Chr.11: 63558382-63558382 on Build GRCh38
rs75523454 Chr.11: 63558316-63558316 on Build GRCh38
rs201347954 Chr.11: 63558410-63558410 on Build GRCh38
rs137992585 Chr.11: 63558440-63558440 on Build GRCh38
rs764173011 Chr.11: 63558431-63558431 on Build GRCh38
rs528540614 Chr.11: 63558311-63558311 on Build GRCh38
rs770618531 Chr.11: 63558490-63558490 on Build GRCh38
rs750975860 Chr.11: 63558455-63558455 on Build GRCh38
rs375507875 Chr.11: 63558497-63558497 on Build GRCh38
rs766419738 Chr.11: 63558402-63558402 on Build GRCh38
rs149474269 Chr.11: 63558472-63558472 on Build GRCh38
rs898116838 Chr.11: 63558351-63558351 on Build GRCh38
rs959427743 Chr.11: 63558310-63558310 on Build GRCh38
rs778420436 Chr.11: 63558335-63558335 on Build GRCh38

View Full Product Details