Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 65537476-65537727 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AXIN2
Entrez Gene ID
8313
Gene Name
axin 2
Gene Aliases
AXIL, ODCRCS
UniGene
Hs.156527
Gene Chromosome Location
Chr.17: 65528563-65561622 on Build GRCh38
Transcripts
NM_004655.3 XM_011525319.2 XM_011525320.1 XM_011525321.2 XM_017025192.1 XM_017025193.1 XM_017025194.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1385328 c.1532C>T p.T511M Chr.17: 65537504-65537504 on Build GRCh38
3820280 c.1437C>T p.S479S Chr.17: 65537599-65537599 on Build GRCh38
2796700 c.1469C>T p.A490V Chr.17: 65537567-65537567 on Build GRCh38
6923994 c.1391C>T p.S464F Chr.17: 65537645-65537645 on Build GRCh38
3890203 c.1466C>T p.P489L Chr.17: 65537570-65537570 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368525111 Chr.17: 65537660-65537660 on Build GRCh38
rs367697282 Chr.17: 65537558-65537558 on Build GRCh38
rs748235355 Chr.17: 65537546-65537547 on Build GRCh38
rs745555433 Chr.17: 65537630-65537630 on Build GRCh38
rs764640447 Chr.17: 65537642-65537642 on Build GRCh38
rs978541876 Chr.17: 65537532-65537532 on Build GRCh38
rs778327226 Chr.17: 65537510-65537510 on Build GRCh38
rs1032366089 Chr.17: 65537657-65537657 on Build GRCh38
rs760282693 Chr.17: 65537638-65537638 on Build GRCh38
rs758495825 Chr.17: 65537591-65537591 on Build GRCh38
rs746454700 Chr.17: 65537680-65537680 on Build GRCh38
rs141014640 Chr.17: 65537506-65537506 on Build GRCh38
rs137933764 Chr.17: 65537600-65537600 on Build GRCh38
rs777699064 Chr.17: 65537619-65537619 on Build GRCh38
rs777283610 Chr.17: 65537645-65537645 on Build GRCh38
rs781185996 Chr.17: 65537540-65537540 on Build GRCh38
rs730881396 Chr.17: 65537555-65537555 on Build GRCh38
rs770054605 Chr.17: 65537615-65537617 on Build GRCh38
rs779863826 Chr.17: 65537673-65537673 on Build GRCh38
rs372513593 Chr.17: 65537553-65537553 on Build GRCh38
rs200883019 Chr.17: 65537504-65537504 on Build GRCh38
rs770556906 Chr.17: 65537551-65537551 on Build GRCh38
rs762233852 Chr.17: 65537646-65537646 on Build GRCh38
rs950007141 Chr.17: 65537612-65537612 on Build GRCh38
rs778612433 Chr.17: 65537667-65537667 on Build GRCh38
rs771579039 Chr.17: 65537635-65537635 on Build GRCh38
rs9914661 Chr.17: 65537653-65537653 on Build GRCh38
rs754079144 Chr.17: 65537525-65537525 on Build GRCh38
rs758592586 Chr.17: 65537651-65537651 on Build GRCh38
rs975950606 Chr.17: 65537683-65537683 on Build GRCh38
rs754204017 Chr.17: 65537577-65537577 on Build GRCh38
rs779029020 Chr.17: 65537534-65537534 on Build GRCh38
rs566313620 Chr.17: 65537656-65537656 on Build GRCh38
rs908388069 Chr.17: 65537636-65537636 on Build GRCh38
rs369864600 Chr.17: 65537698-65537698 on Build GRCh38
rs931608809 Chr.17: 65537560-65537560 on Build GRCh38
rs570443161 Chr.17: 65537614-65537615 on Build GRCh38
rs758413474 Chr.17: 65537537-65537537 on Build GRCh38
rs777265234 Chr.17: 65537687-65537687 on Build GRCh38
rs1028974721 Chr.17: 65537501-65537501 on Build GRCh38
rs764728243 Chr.17: 65537571-65537571 on Build GRCh38
rs757512425 Chr.17: 65537526-65537526 on Build GRCh38
rs372841853 Chr.17: 65537668-65537668 on Build GRCh38
rs9915936 Chr.17: 65537671-65537671 on Build GRCh38
rs878854721 Chr.17: 65537647-65537647 on Build GRCh38
rs745903369 Chr.17: 65537536-65537536 on Build GRCh38
rs765845602 Chr.17: 65537503-65537503 on Build GRCh38
rs765434564 Chr.17: 65537587-65537587 on Build GRCh38
rs748049932 Chr.17: 65537614-65537614 on Build GRCh38
rs367938045 Chr.17: 65537507-65537507 on Build GRCh38
rs376936740 Chr.17: 65537505-65537505 on Build GRCh38
rs749012756 Chr.17: 65537621-65537621 on Build GRCh38
rs753827256 Chr.17: 65537664-65537664 on Build GRCh38
rs730881395 Chr.17: 65537649-65537649 on Build GRCh38
rs530465133 Chr.17: 65537588-65537588 on Build GRCh38
rs773033390 Chr.17: 65537550-65537550 on Build GRCh38
rs750678891 Chr.17: 65537590-65537590 on Build GRCh38
rs777680532 Chr.17: 65537661-65537661 on Build GRCh38
rs762279806 Chr.17: 65537689-65537689 on Build GRCh38
rs149483825 Chr.17: 65537626-65537626 on Build GRCh38
rs976214591 Chr.17: 65537539-65537539 on Build GRCh38
rs757050940 Chr.17: 65537665-65537665 on Build GRCh38
rs868617060 Chr.17: 65537702-65537702 on Build GRCh38
rs544255284 Chr.17: 65537625-65537625 on Build GRCh38
rs1133683 Chr.17: 65537650-65537650 on Build GRCh38
rs769558841 Chr.17: 65537546-65537546 on Build GRCh38
rs761303245 Chr.17: 65537565-65537565 on Build GRCh38
rs886041137 Chr.17: 65537643-65537643 on Build GRCh38
rs1048751630 Chr.17: 65537563-65537563 on Build GRCh38
rs756008626 Chr.17: 65537615-65537615 on Build GRCh38
rs768349653 Chr.17: 65537684-65537684 on Build GRCh38
rs150292240 Chr.17: 65537545-65537545 on Build GRCh38
rs371446806 Chr.17: 65537586-65537586 on Build GRCh38
rs886053272 Chr.17: 65537593-65537593 on Build GRCh38

View Full Product Details