Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 61416230-61416497 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPSF7
Entrez Gene ID
79869
Gene Name
cleavage and polyadenylation specific factor 7
Gene Aliases
CFIm59
UniGene
Hs.718984
Gene Chromosome Location
Chr.11: 61402648-61429992 on Build GRCh38
Transcripts
NM_001136040.2 NM_001142565.1 NM_024811.3 XM_005274298.4 XM_005274299.4 XM_005274303.4 XM_011545257.2 XM_011545258.2 XM_011545259.2 XM_011545260.2 XM_011545261.2 XM_011545262.2 XM_011545263.2 XM_017018345.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4674478 c.806delC p.P269fs*43 Chr.11: 61416264-61416264 on Build GRCh38
1704118 c.763C>T p.P255S Chr.11: 61416307-61416307 on Build GRCh38
6213938 c.846A>C p.P282P Chr.11: 61416353-61416353 on Build GRCh38
6069705 c.806A>G p.N269S Chr.11: 61416393-61416393 on Build GRCh38
7269465 c.819G>A p.S273S Chr.11: 61416380-61416380 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755313692 Chr.11: 61416293-61416295 on Build GRCh38
rs774144889 Chr.11: 61416378-61416378 on Build GRCh38
rs750933170 Chr.11: 61416262-61416262 on Build GRCh38
rs925568523 Chr.11: 61416271-61416271 on Build GRCh38
rs758903187 Chr.11: 61416269-61416269 on Build GRCh38
rs770109461 Chr.11: 61416292-61416293 on Build GRCh38
rs781320888 Chr.11: 61416455-61416455 on Build GRCh38
rs374077859 Chr.11: 61416414-61416414 on Build GRCh38
rs1041606507 Chr.11: 61416448-61416448 on Build GRCh38
rs781438426 Chr.11: 61416341-61416341 on Build GRCh38
rs201389052 Chr.11: 61416393-61416393 on Build GRCh38
rs1018121942 Chr.11: 61416427-61416427 on Build GRCh38
rs548830763 Chr.11: 61416413-61416413 on Build GRCh38
rs749218250 Chr.11: 61416370-61416370 on Build GRCh38
rs748218062 Chr.11: 61416346-61416346 on Build GRCh38
rs557273852 Chr.11: 61416289-61416289 on Build GRCh38
rs758991309 Chr.11: 61416429-61416429 on Build GRCh38
rs371227588 Chr.11: 61416404-61416404 on Build GRCh38
rs768786067 Chr.11: 61416333-61416333 on Build GRCh38
rs552252814 Chr.11: 61416425-61416425 on Build GRCh38
rs752006554 Chr.11: 61416430-61416430 on Build GRCh38
rs369162186 Chr.11: 61416353-61416353 on Build GRCh38
rs748388788 Chr.11: 61416462-61416462 on Build GRCh38
rs532521319 Chr.11: 61416382-61416382 on Build GRCh38
rs767105394 Chr.11: 61416385-61416385 on Build GRCh38
rs944294721 Chr.11: 61416447-61416447 on Build GRCh38
rs770786938 Chr.11: 61416376-61416376 on Build GRCh38
rs201985915 Chr.11: 61416469-61416469 on Build GRCh38
rs374683197 Chr.11: 61416301-61416301 on Build GRCh38
rs199830766 Chr.11: 61416329-61416329 on Build GRCh38
rs779324227 Chr.11: 61416259-61416259 on Build GRCh38
rs904328304 Chr.11: 61416380-61416380 on Build GRCh38
rs756216467 Chr.11: 61416468-61416468 on Build GRCh38
rs267603065 Chr.11: 61416348-61416348 on Build GRCh38
rs373307890 Chr.11: 61416390-61416390 on Build GRCh38
rs148252018 Chr.11: 61416356-61416356 on Build GRCh38
rs931652189 Chr.11: 61416359-61416359 on Build GRCh38
rs755339835 Chr.11: 61416444-61416444 on Build GRCh38

View Full Product Details