Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 99359512-99359773 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARPC1A
Entrez Gene ID
10552
Gene Name
actin related protein 2/3 complex subunit 1A
Gene Aliases
Arc40, HEL-68, HEL-S-307, SOP2Hs, SOP2L
UniGene
Hs.124126
Gene Chromosome Location
Chr.7: 99325873-99366262 on Build GRCh38
Transcripts
NM_001190996.1 NM_006409.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1453252 c.896C>T p.A299V Chr.7: 99359651-99359651 on Build GRCh38
4663478 c.904C>T p.R302C Chr.7: 99359659-99359659 on Build GRCh38
6605107 c.944G>A p.R315H Chr.7: 99359699-99359699 on Build GRCh38
4663481 c.810G>A p.T270T Chr.7: 99359706-99359706 on Build GRCh38
1453251 c.795T>C p.H265H Chr.7: 99359550-99359550 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200755413 Chr.7: 99359592-99359592 on Build GRCh38
rs142247800 Chr.7: 99359563-99359563 on Build GRCh38
rs755659135 Chr.7: 99359617-99359617 on Build GRCh38
rs771914193 Chr.7: 99359754-99359754 on Build GRCh38
rs199843941 Chr.7: 99359748-99359748 on Build GRCh38
rs755391152 Chr.7: 99359704-99359704 on Build GRCh38
rs901011710 Chr.7: 99359558-99359558 on Build GRCh38
rs774352467 Chr.7: 99359538-99359538 on Build GRCh38
rs148830024 Chr.7: 99359752-99359752 on Build GRCh38
rs749230685 Chr.7: 99359634-99359634 on Build GRCh38
rs2286160 Chr.7: 99359602-99359602 on Build GRCh38
rs748858384 Chr.7: 99359736-99359736 on Build GRCh38
rs529951024 Chr.7: 99359638-99359638 on Build GRCh38
rs748107828 Chr.7: 99359538-99359539 on Build GRCh38
rs767015647 Chr.7: 99359705-99359705 on Build GRCh38
rs368731088 Chr.7: 99359747-99359747 on Build GRCh38
rs143044515 Chr.7: 99359586-99359586 on Build GRCh38
rs150324163 Chr.7: 99359664-99359664 on Build GRCh38
rs752395773 Chr.7: 99359706-99359706 on Build GRCh38
rs141228867 Chr.7: 99359652-99359652 on Build GRCh38
rs576410572 Chr.7: 99359587-99359587 on Build GRCh38
rs200959894 Chr.7: 99359585-99359585 on Build GRCh38
rs754555024 Chr.7: 99359601-99359601 on Build GRCh38
rs774268688 Chr.7: 99359640-99359640 on Build GRCh38
rs922447965 Chr.7: 99359557-99359557 on Build GRCh38
rs576105816 Chr.7: 99359577-99359577 on Build GRCh38
rs773348651 Chr.7: 99359699-99359699 on Build GRCh38
rs765365489 Chr.7: 99359685-99359685 on Build GRCh38
rs776934695 Chr.7: 99359669-99359669 on Build GRCh38
rs781337932 Chr.7: 99359579-99359579 on Build GRCh38
rs373536333 Chr.7: 99359537-99359537 on Build GRCh38
rs747559622 Chr.7: 99359611-99359611 on Build GRCh38
rs910006701 Chr.7: 99359616-99359616 on Build GRCh38
rs763426021 Chr.7: 99359700-99359700 on Build GRCh38
rs1014475489 Chr.7: 99359746-99359746 on Build GRCh38
rs777784312 Chr.7: 99359627-99359627 on Build GRCh38
rs137914882 Chr.7: 99359678-99359678 on Build GRCh38
rs780382694 Chr.7: 99359598-99359598 on Build GRCh38
rs373720602 Chr.7: 99359718-99359718 on Build GRCh38
rs766909995 Chr.7: 99359701-99359701 on Build GRCh38
rs751880782 Chr.7: 99359703-99359703 on Build GRCh38
rs187071817 Chr.7: 99359584-99359584 on Build GRCh38

View Full Product Details