Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 168358180-168358407 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX60L
Entrez Gene ID
91351
Gene Name
DEAD-box helicase 60-like
Gene Aliases
-
UniGene
Hs.535011
Gene Chromosome Location
Chr.4: 168356735-168480514 on Build GRCh38
Transcripts
NM_001012967.2 XM_005263341.4 XM_005263342.2 XM_005263343.2 XM_005263345.4 XM_006714408.2 XM_011532402.2 XM_011532403.2 XM_011532404.1 XM_011532405.1 XM_011532406.1 XM_011532409.2 XM_017008829.1 XM_017008830.1 XM_017008831.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3335990 c.5068T>G p.Y1690D Chr.4: 168358200-168358200 on Build GRCh38
7215809 c.5071G>A p.E1691K Chr.4: 168358197-168358197 on Build GRCh38
3409160 c.5024G>A p.R1675H Chr.4: 168358244-168358244 on Build GRCh38
6167077 c.5024G>C p.R1675P Chr.4: 168358244-168358244 on Build GRCh38
7243958 c.5055G>A p.L1685L Chr.4: 168358213-168358213 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759068019 Chr.4: 168358246-168358246 on Build GRCh38
rs770079691 Chr.4: 168358231-168358231 on Build GRCh38
rs759180268 Chr.4: 168358289-168358289 on Build GRCh38
rs115559281 Chr.4: 168358366-168358366 on Build GRCh38
rs1049979114 Chr.4: 168358258-168358258 on Build GRCh38
rs745338983 Chr.4: 168358288-168358290 on Build GRCh38
rs951935974 Chr.4: 168358316-168358316 on Build GRCh38
rs760415358 Chr.4: 168358260-168358260 on Build GRCh38
rs1024150728 Chr.4: 168358328-168358328 on Build GRCh38
rs964053425 Chr.4: 168358254-168358254 on Build GRCh38
rs769991271 Chr.4: 168358310-168358310 on Build GRCh38
rs753677644 Chr.4: 168358284-168358289 on Build GRCh38
rs1032862168 Chr.4: 168358352-168358352 on Build GRCh38
rs778690370 Chr.4: 168358284-168358285 on Build GRCh38
rs766664760 Chr.4: 168358265-168358265 on Build GRCh38
rs748313549 Chr.4: 168358304-168358304 on Build GRCh38
rs752357025 Chr.4: 168358256-168358256 on Build GRCh38
rs200274079 Chr.4: 168358249-168358249 on Build GRCh38
rs748397533 Chr.4: 168358228-168358228 on Build GRCh38
rs763367090 Chr.4: 168358238-168358238 on Build GRCh38
rs11727892 Chr.4: 168358287-168358287 on Build GRCh38
rs1001425172 Chr.4: 168358347-168358347 on Build GRCh38
rs778037300 Chr.4: 168358319-168358319 on Build GRCh38
rs771464614 Chr.4: 168358244-168358244 on Build GRCh38
rs371572312 Chr.4: 168358245-168358245 on Build GRCh38
rs989424913 Chr.4: 168358362-168358363 on Build GRCh38
rs781134168 Chr.4: 168358301-168358301 on Build GRCh38
rs907779110 Chr.4: 168358361-168358361 on Build GRCh38
rs778886855 Chr.4: 168358286-168358286 on Build GRCh38
rs748612096 Chr.4: 168358306-168358306 on Build GRCh38
rs11736382 Chr.4: 168358213-168358213 on Build GRCh38
rs77455438 Chr.4: 168358353-168358353 on Build GRCh38
rs757153520 Chr.4: 168358284-168358284 on Build GRCh38
rs970257337 Chr.4: 168358334-168358335 on Build GRCh38
rs779560346 Chr.4: 168358299-168358301 on Build GRCh38
rs771660670 Chr.4: 168358298-168358299 on Build GRCh38
rs777647425 Chr.4: 168358281-168358284 on Build GRCh38
rs756843567 Chr.4: 168358285-168358287 on Build GRCh38
rs753677260 Chr.4: 168358276-168358276 on Build GRCh38
rs755043596 Chr.4: 168358292-168358292 on Build GRCh38
rs140243454 Chr.4: 168358236-168358236 on Build GRCh38

View Full Product Details