Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 73395929-73396200 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANXA7
Entrez Gene ID
310
Gene Name
annexin A7
Gene Aliases
ANX7, SNX, SYNEXIN
UniGene
Hs.631827
Gene Chromosome Location
Chr.10: 73375101-73414085 on Build GRCh38
Transcripts
NM_001156.4 NM_001320879.1 NM_001320880.1 NM_004034.3 XM_017016162.1 XM_017016163.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
465943 c.482C>T p.S161F Chr.10: 73396063-73396063 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs755690290 Chr.10: 73396075-73396075 on Build GRCh38
rs1039533855 Chr.10: 73396131-73396131 on Build GRCh38
rs374662699 Chr.10: 73396143-73396143 on Build GRCh38
rs767683784 Chr.10: 73396038-73396041 on Build GRCh38
rs771054250 Chr.10: 73396088-73396088 on Build GRCh38
rs1053057608 Chr.10: 73396171-73396171 on Build GRCh38
rs182423992 Chr.10: 73396033-73396033 on Build GRCh38
rs373680861 Chr.10: 73396034-73396034 on Build GRCh38
rs776544921 Chr.10: 73396103-73396103 on Build GRCh38
rs759075802 Chr.10: 73396156-73396156 on Build GRCh38
rs375002246 Chr.10: 73396110-73396110 on Build GRCh38
rs1024080126 Chr.10: 73395964-73395964 on Build GRCh38
rs574638555 Chr.10: 73395974-73395974 on Build GRCh38
rs901118437 Chr.10: 73396132-73396132 on Build GRCh38
rs767644875 Chr.10: 73396018-73396018 on Build GRCh38
rs776869039 Chr.10: 73396011-73396011 on Build GRCh38
rs367821209 Chr.10: 73396090-73396090 on Build GRCh38
rs754426993 Chr.10: 73396053-73396053 on Build GRCh38
rs567451322 Chr.10: 73396125-73396125 on Build GRCh38
rs905635461 Chr.10: 73395984-73395984 on Build GRCh38
rs749075182 Chr.10: 73396086-73396086 on Build GRCh38
rs146403699 Chr.10: 73396146-73396146 on Build GRCh38
rs371572648 Chr.10: 73396096-73396096 on Build GRCh38
rs935295721 Chr.10: 73396159-73396159 on Build GRCh38
rs528513232 Chr.10: 73396049-73396049 on Build GRCh38
rs768745097 Chr.10: 73396102-73396102 on Build GRCh38
rs750680594 Chr.10: 73396104-73396105 on Build GRCh38
rs763511659 Chr.10: 73396041-73396041 on Build GRCh38
rs760509837 Chr.10: 73396109-73396110 on Build GRCh38
rs371507882 Chr.10: 73395993-73395993 on Build GRCh38
rs1007137162 Chr.10: 73396130-73396130 on Build GRCh38
rs980930843 Chr.10: 73395972-73395972 on Build GRCh38
rs7097105 Chr.10: 73395960-73395960 on Build GRCh38
rs763237004 Chr.10: 73396128-73396128 on Build GRCh38
rs948233877 Chr.10: 73395961-73395961 on Build GRCh38
rs922083062 Chr.10: 73396105-73396105 on Build GRCh38
rs762145780 Chr.10: 73396107-73396107 on Build GRCh38
rs149924138 Chr.10: 73396067-73396067 on Build GRCh38
rs762118234 Chr.10: 73396019-73396019 on Build GRCh38
rs765676869 Chr.10: 73396027-73396027 on Build GRCh38
rs989171718 Chr.10: 73395958-73395958 on Build GRCh38
rs766284993 Chr.10: 73396112-73396113 on Build GRCh38
rs777669484 Chr.10: 73396085-73396085 on Build GRCh38
rs753773232 Chr.10: 73396133-73396140 on Build GRCh38
rs1027356279 Chr.10: 73396045-73396045 on Build GRCh38
rs145770891 Chr.10: 73396069-73396069 on Build GRCh38

View Full Product Details