Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 44926541-44926814 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF2B3
Entrez Gene ID
8891
Gene Name
eukaryotic translation initiation factor 2B subunit gamma
Gene Aliases
EIF-2B, EIF2Bgamma
UniGene
Hs.533549
Gene Chromosome Location
Chr.1: 44850522-44986722 on Build GRCh38
Transcripts
NM_001166588.2 NM_001261418.1 NM_020365.4 XM_017002745.1 XM_017002746.1 XM_017002747.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3490183 c.550G>A p.G184R Chr.1: 44926644-44926644 on Build GRCh38
4008456 c.517G>A p.E173K Chr.1: 44926677-44926677 on Build GRCh38
910096 c.535G>T p.E179* Chr.1: 44926659-44926659 on Build GRCh38
4008457 c.473T>G p.I158S Chr.1: 44926721-44926721 on Build GRCh38
6652715 c.508A>G p.M170V Chr.1: 44926686-44926686 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs112906939 Chr.1: 44926659-44926659 on Build GRCh38
rs770951184 Chr.1: 44926599-44926599 on Build GRCh38
rs898762337 Chr.1: 44926657-44926657 on Build GRCh38
rs570952040 Chr.1: 44926756-44926756 on Build GRCh38
rs761940558 Chr.1: 44926581-44926581 on Build GRCh38
rs776973264 Chr.1: 44926580-44926580 on Build GRCh38
rs758794576 Chr.1: 44926755-44926756 on Build GRCh38
rs560755503 Chr.1: 44926774-44926774 on Build GRCh38
rs751321524 Chr.1: 44926636-44926636 on Build GRCh38
rs547357260 Chr.1: 44926755-44926755 on Build GRCh38
rs780554112 Chr.1: 44926735-44926735 on Build GRCh38
rs796116349 Chr.1: 44926756-44926757 on Build GRCh38
rs902550651 Chr.1: 44926571-44926572 on Build GRCh38
rs201318616 Chr.1: 44926683-44926683 on Build GRCh38
rs960112442 Chr.1: 44926720-44926720 on Build GRCh38
rs765344307 Chr.1: 44926582-44926582 on Build GRCh38
rs147773599 Chr.1: 44926730-44926730 on Build GRCh38
rs999158795 Chr.1: 44926615-44926615 on Build GRCh38
rs775973608 Chr.1: 44926638-44926638 on Build GRCh38
rs748387340 Chr.1: 44926780-44926780 on Build GRCh38
rs527440686 Chr.1: 44926750-44926750 on Build GRCh38
rs755805377 Chr.1: 44926728-44926728 on Build GRCh38
rs773223478 Chr.1: 44926589-44926589 on Build GRCh38
rs769958739 Chr.1: 44926782-44926782 on Build GRCh38
rs1022637609 Chr.1: 44926746-44926746 on Build GRCh38
rs767209968 Chr.1: 44926692-44926692 on Build GRCh38
rs762904394 Chr.1: 44926598-44926598 on Build GRCh38
rs534753916 Chr.1: 44926756-44926757 on Build GRCh38
rs752434700 Chr.1: 44926697-44926697 on Build GRCh38
rs545295324 Chr.1: 44926630-44926630 on Build GRCh38
rs769135301 Chr.1: 44926773-44926773 on Build GRCh38
rs754858644 Chr.1: 44926642-44926642 on Build GRCh38
rs769156320 Chr.1: 44926767-44926767 on Build GRCh38
rs564587689 Chr.1: 44926731-44926731 on Build GRCh38

View Full Product Details