Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 304499-304772 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUSP22
Entrez Gene ID
56940
Gene Name
dual specificity phosphatase 22
Gene Aliases
JKAP, JSP-1, JSP1, LMW-DSP2, LMWDSP2, MKP-x, MKPX, VHX
UniGene
Hs.29106
Gene Chromosome Location
Chr.6: 292057-351355 on Build GRCh38
Transcripts
NM_001286555.1 NM_020185.4 NR_104473.1 NR_104474.1 NR_104475.1 XM_017011061.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3251332 c.39C>A p.Y13* Chr.6: 304645-304645 on Build GRCh38
5357598 c.22-1G>T p.? Chr.6: 304627-304627 on Build GRCh38
3251333 c.40A>G p.I14V Chr.6: 304646-304646 on Build GRCh38
3624251 c.32G>A p.G11D Chr.6: 304638-304638 on Build GRCh38
6350700 c.43G>C p.G15R Chr.6: 304649-304649 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs557309877 Chr.6: 304597-304597 on Build GRCh38
rs769701077 Chr.6: 304590-304590 on Build GRCh38
rs567730576 Chr.6: 304531-304531 on Build GRCh38
rs967656875 Chr.6: 304591-304591 on Build GRCh38
rs773445715 Chr.6: 304586-304587 on Build GRCh38
rs752294946 Chr.6: 304616-304619 on Build GRCh38
rs373379863 Chr.6: 304577-304577 on Build GRCh38
rs767041471 Chr.6: 304639-304639 on Build GRCh38
rs781191751 Chr.6: 304621-304621 on Build GRCh38
rs773158137 Chr.6: 304592-304592 on Build GRCh38
rs777829242 Chr.6: 304664-304664 on Build GRCh38
rs768667760 Chr.6: 304584-304584 on Build GRCh38
rs781270005 Chr.6: 304653-304653 on Build GRCh38
rs745467173 Chr.6: 304696-304696 on Build GRCh38
rs565428090 Chr.6: 304578-304578 on Build GRCh38
rs768196303 Chr.6: 304710-304710 on Build GRCh38
rs142889998 Chr.6: 304636-304636 on Build GRCh38
rs1038712257 Chr.6: 304689-304689 on Build GRCh38
rs766974988 Chr.6: 304605-304605 on Build GRCh38
rs151205644 Chr.6: 304637-304637 on Build GRCh38
rs755084477 Chr.6: 304620-304620 on Build GRCh38
rs756063227 Chr.6: 304657-304657 on Build GRCh38
rs979935752 Chr.6: 304518-304518 on Build GRCh38
rs748265668 Chr.6: 304655-304655 on Build GRCh38
rs762582315 Chr.6: 304681-304683 on Build GRCh38
rs751795600 Chr.6: 304651-304651 on Build GRCh38
rs771627323 Chr.6: 304700-304700 on Build GRCh38
rs376352810 Chr.6: 304582-304582 on Build GRCh38
rs765755658 Chr.6: 304645-304645 on Build GRCh38
rs761323622 Chr.6: 304608-304609 on Build GRCh38
rs996586626 Chr.6: 304544-304544 on Build GRCh38
rs1029373692 Chr.6: 304720-304720 on Build GRCh38
rs368899941 Chr.6: 304574-304574 on Build GRCh38
rs760156834 Chr.6: 304705-304705 on Build GRCh38
rs774070772 Chr.6: 304692-304692 on Build GRCh38
rs925997103 Chr.6: 304614-304614 on Build GRCh38
rs201434867 Chr.6: 304646-304646 on Build GRCh38
rs535538272 Chr.6: 304540-304540 on Build GRCh38
rs766971540 Chr.6: 304616-304617 on Build GRCh38
rs780069821 Chr.6: 304579-304579 on Build GRCh38
rs762921083 Chr.6: 304648-304648 on Build GRCh38
rs754364554 Chr.6: 304607-304607 on Build GRCh38
rs755173245 Chr.6: 304652-304652 on Build GRCh38
rs762625031 Chr.6: 304594-304594 on Build GRCh38
rs759061678 Chr.6: 304602-304602 on Build GRCh38
rs957430410 Chr.6: 304622-304622 on Build GRCh38
rs375630773 Chr.6: 304688-304688 on Build GRCh38
rs757817208 Chr.6: 304609-304609 on Build GRCh38
rs766724185 Chr.6: 304649-304649 on Build GRCh38
rs371110899 Chr.6: 304588-304588 on Build GRCh38
rs750819088 Chr.6: 304613-304613 on Build GRCh38
rs773682433 Chr.6: 304598-304598 on Build GRCh38
rs776604326 Chr.6: 304585-304585 on Build GRCh38
rs758572784 Chr.6: 304618-304618 on Build GRCh38
rs762268962 Chr.6: 304644-304644 on Build GRCh38
rs749115010 Chr.6: 304633-304633 on Build GRCh38
rs774875404 Chr.6: 304640-304640 on Build GRCh38
rs775017735 Chr.6: 304703-304703 on Build GRCh38
rs773099083 Chr.6: 304629-304629 on Build GRCh38
rs769861594 Chr.6: 304628-304628 on Build GRCh38
rs780233657 Chr.6: 304619-304619 on Build GRCh38
rs1016628929 Chr.6: 304532-304532 on Build GRCh38
rs1005170267 Chr.6: 304527-304527 on Build GRCh38

View Full Product Details