Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 9116683-9116940 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C16orf72
Entrez Gene ID
29035
Gene Name
chromosome 16 open reading frame 72
Gene Aliases
PRO0149
UniGene
Hs.221497
Gene Chromosome Location
Chr.16: 9091329-9119698 on Build GRCh38
Transcripts
NM_014117.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2924125 c.715T>A p.S239T Chr.16: 9116799-9116799 on Build GRCh38
2924124 c.658T>C p.S220P Chr.16: 9116742-9116742 on Build GRCh38
1493792 c.611G>A p.S204N Chr.16: 9116695-9116695 on Build GRCh38
1380643 c.811C>T p.R271C Chr.16: 9116895-9116895 on Build GRCh38
1380642 c.697G>A p.D233N Chr.16: 9116781-9116781 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756286069 Chr.16: 9116917-9116917 on Build GRCh38
rs750786253 Chr.16: 9116858-9116858 on Build GRCh38
rs774003402 Chr.16: 9116790-9116790 on Build GRCh38
rs775616262 Chr.16: 9116814-9116814 on Build GRCh38
rs982440425 Chr.16: 9116705-9116705 on Build GRCh38
rs776209991 Chr.16: 9116830-9116830 on Build GRCh38
rs760712474 Chr.16: 9116817-9116817 on Build GRCh38
rs1023428811 Chr.16: 9116916-9116916 on Build GRCh38
rs763474218 Chr.16: 9116827-9116827 on Build GRCh38
rs749850275 Chr.16: 9116907-9116907 on Build GRCh38
rs999506590 Chr.16: 9116871-9116871 on Build GRCh38
rs765001709 Chr.16: 9116853-9116853 on Build GRCh38
rs766852526 Chr.16: 9116873-9116873 on Build GRCh38
rs942484018 Chr.16: 9116754-9116754 on Build GRCh38
rs542361654 Chr.16: 9116919-9116919 on Build GRCh38
rs745723920 Chr.16: 9116792-9116792 on Build GRCh38
rs781070790 Chr.16: 9116895-9116895 on Build GRCh38
rs142730369 Chr.16: 9116849-9116849 on Build GRCh38
rs372524467 Chr.16: 9116744-9116744 on Build GRCh38
rs781361030 Chr.16: 9116736-9116736 on Build GRCh38
rs748317124 Chr.16: 9116897-9116897 on Build GRCh38
rs1039176829 Chr.16: 9116781-9116781 on Build GRCh38
rs778320406 Chr.16: 9116859-9116859 on Build GRCh38
rs148092798 Chr.16: 9116801-9116801 on Build GRCh38
rs755477383 Chr.16: 9116894-9116894 on Build GRCh38
rs997742685 Chr.16: 9116836-9116836 on Build GRCh38
rs900688500 Chr.16: 9116825-9116825 on Build GRCh38
rs34877028 Chr.16: 9116853-9116854 on Build GRCh38
rs770064445 Chr.16: 9116764-9116764 on Build GRCh38
rs752018395 Chr.16: 9116874-9116874 on Build GRCh38
rs200766200 Chr.16: 9116731-9116731 on Build GRCh38
rs754999038 Chr.16: 9116732-9116732 on Build GRCh38
rs34869458 Chr.16: 9116746-9116746 on Build GRCh38

View Full Product Details