Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 85596281-85596532 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGTPBP1
Entrez Gene ID
23287
Gene Name
ATP/GTP binding protein 1
Gene Aliases
CCP1, NNA1
UniGene
Hs.719980
Gene Chromosome Location
Chr.9: 85543799-85742029 on Build GRCh38
Transcripts
NM_001286715.1 NM_001286717.1 NM_015239.2 XM_005251847.3 XM_005251848.2 XM_005251849.3 XM_011518418.2 XM_011518420.2 XM_017014544.1 XM_017014545.1 XM_017014546.1 XM_017014547.1 XM_017014548.1 XM_017014549.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7196886 c.2278A>G p.M760V Chr.9: 85596387-85596387 on Build GRCh38
3675400 c.2229C>A p.L743L Chr.9: 85596436-85596436 on Build GRCh38
5024619 c.2423A>C p.K808T Chr.9: 85596362-85596362 on Build GRCh38
7275622 c.2399T>C p.M800T Chr.9: 85596386-85596386 on Build GRCh38
4813246 c.2401G>T p.G801W Chr.9: 85596384-85596384 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200123422 Chr.9: 85596426-85596426 on Build GRCh38
rs771822098 Chr.9: 85596469-85596470 on Build GRCh38
rs752823752 Chr.9: 85596318-85596318 on Build GRCh38
rs753199638 Chr.9: 85596485-85596485 on Build GRCh38
rs768516291 Chr.9: 85596403-85596403 on Build GRCh38
rs988266420 Chr.9: 85596346-85596346 on Build GRCh38
rs372210443 Chr.9: 85596324-85596324 on Build GRCh38
rs771912159 Chr.9: 85596365-85596365 on Build GRCh38
rs112063816 Chr.9: 85596448-85596448 on Build GRCh38
rs766800584 Chr.9: 85596468-85596468 on Build GRCh38
rs781644979 Chr.9: 85596315-85596315 on Build GRCh38
rs746997311 Chr.9: 85596397-85596397 on Build GRCh38
rs371844488 Chr.9: 85596486-85596486 on Build GRCh38
rs779664281 Chr.9: 85596389-85596389 on Build GRCh38
rs778478854 Chr.9: 85596455-85596466 on Build GRCh38
rs372848034 Chr.9: 85596435-85596435 on Build GRCh38
rs777769474 Chr.9: 85596497-85596497 on Build GRCh38
rs76507545 Chr.9: 85596454-85596454 on Build GRCh38
rs747785622 Chr.9: 85596462-85596463 on Build GRCh38
rs41282441 Chr.9: 85596472-85596472 on Build GRCh38
rs12345317 Chr.9: 85596331-85596331 on Build GRCh38
rs759922566 Chr.9: 85596476-85596476 on Build GRCh38
rs377196703 Chr.9: 85596460-85596460 on Build GRCh38
rs535786336 Chr.9: 85596467-85596467 on Build GRCh38
rs747715117 Chr.9: 85596427-85596427 on Build GRCh38
rs868705257 Chr.9: 85596364-85596364 on Build GRCh38
rs768071362 Chr.9: 85596479-85596479 on Build GRCh38

View Full Product Details