Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 169596434-169596659 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPDL1
Entrez Gene ID
54908
Gene Name
spindle apparatus coiled-coil protein 1
Gene Aliases
CCDC99
UniGene
Hs.368710
Gene Chromosome Location
Chr.5: 169583634-169604778 on Build GRCh38
Transcripts
NM_017785.4 XM_011534585.1 XM_011534586.2 XM_011534587.2 XM_011534588.2 XM_011534589.2 XM_011534590.2 XM_017009601.1 XM_017009602.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
736812 c.948G>T p.Q316H Chr.5: 169596617-169596617 on Build GRCh38
402742 c.970G>T p.E324* Chr.5: 169596639-169596639 on Build GRCh38
6102963 c.936A>G p.E312E Chr.5: 169596605-169596605 on Build GRCh38
4935825 c.932C>T p.T311I Chr.5: 169596601-169596601 on Build GRCh38
3994175 c.941A>G p.E314G Chr.5: 169596610-169596610 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753614593 Chr.5: 169596574-169596574 on Build GRCh38
rs892207403 Chr.5: 169596470-169596470 on Build GRCh38
rs758172358 Chr.5: 169596599-169596599 on Build GRCh38
rs752698109 Chr.5: 169596523-169596526 on Build GRCh38
rs774675884 Chr.5: 169596543-169596543 on Build GRCh38
rs753195291 Chr.5: 169596566-169596566 on Build GRCh38
rs377594891 Chr.5: 169596619-169596619 on Build GRCh38
rs1045819571 Chr.5: 169596512-169596512 on Build GRCh38
rs763156009 Chr.5: 169596540-169596540 on Build GRCh38
rs906342095 Chr.5: 169596499-169596499 on Build GRCh38
rs913428277 Chr.5: 169596498-169596498 on Build GRCh38
rs149818046 Chr.5: 169596570-169596570 on Build GRCh38
rs370344964 Chr.5: 169596581-169596581 on Build GRCh38
rs12521871 Chr.5: 169596468-169596468 on Build GRCh38
rs187163357 Chr.5: 169596489-169596489 on Build GRCh38
rs201145666 Chr.5: 169596585-169596585 on Build GRCh38
rs779879065 Chr.5: 169596618-169596618 on Build GRCh38
rs761585669 Chr.5: 169596517-169596520 on Build GRCh38
rs766787065 Chr.5: 169596541-169596541 on Build GRCh38
rs142021941 Chr.5: 169596621-169596621 on Build GRCh38
rs1002125939 Chr.5: 169596505-169596505 on Build GRCh38
rs767936181 Chr.5: 169596565-169596565 on Build GRCh38
rs1009425984 Chr.5: 169596488-169596488 on Build GRCh38
rs750294136 Chr.5: 169596602-169596603 on Build GRCh38
rs145815866 Chr.5: 169596601-169596601 on Build GRCh38
rs773264345 Chr.5: 169596633-169596633 on Build GRCh38
rs766145499 Chr.5: 169596575-169596575 on Build GRCh38
rs140005442 Chr.5: 169596559-169596559 on Build GRCh38

View Full Product Details