Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 130924328-130924567 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FIBCD1
Entrez Gene ID
84929
Gene Name
fibrinogen C domain containing 1
Gene Aliases
-
UniGene
Hs.133205
Gene Chromosome Location
Chr.9: 130902438-130939068 on Build GRCh38
Transcripts
NM_001145106.1 NM_032843.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4966502 c.609C>T p.I203I Chr.9: 130924340-130924340 on Build GRCh38
4553370 c.588G>T p.L196L Chr.9: 130924361-130924361 on Build GRCh38
6386742 c.559T>C p.S187P Chr.9: 130924390-130924390 on Build GRCh38
3847858 c.577A>G p.M193V Chr.9: 130924372-130924372 on Build GRCh38
3654937 c.591G>A p.V197V Chr.9: 130924358-130924358 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs779262160 Chr.9: 130924407-130924407 on Build GRCh38
rs141420720 Chr.9: 130924346-130924346 on Build GRCh38
rs760873952 Chr.9: 130924432-130924432 on Build GRCh38
rs200302172 Chr.9: 130924422-130924422 on Build GRCh38
rs750388347 Chr.9: 130924387-130924388 on Build GRCh38
rs569652526 Chr.9: 130924454-130924454 on Build GRCh38
rs920237633 Chr.9: 130924498-130924498 on Build GRCh38
rs909287961 Chr.9: 130924449-130924449 on Build GRCh38
rs377395597 Chr.9: 130924402-130924402 on Build GRCh38
rs538526443 Chr.9: 130924530-130924530 on Build GRCh38
rs949359958 Chr.9: 130924510-130924510 on Build GRCh38
rs545646071 Chr.9: 130924373-130924373 on Build GRCh38
rs987284690 Chr.9: 130924447-130924447 on Build GRCh38
rs767875793 Chr.9: 130924429-130924429 on Build GRCh38
rs773273935 Chr.9: 130924430-130924430 on Build GRCh38
rs769160164 Chr.9: 130924415-130924415 on Build GRCh38
rs527825208 Chr.9: 130924410-130924410 on Build GRCh38
rs753897161 Chr.9: 130924443-130924443 on Build GRCh38
rs775566308 Chr.9: 130924419-130924419 on Build GRCh38
rs756076743 Chr.9: 130924440-130924441 on Build GRCh38
rs771845696 Chr.9: 130924459-130924459 on Build GRCh38
rs767783274 Chr.9: 130924372-130924372 on Build GRCh38
rs751891931 Chr.9: 130924361-130924361 on Build GRCh38
rs375652381 Chr.9: 130924426-130924426 on Build GRCh38
rs974432704 Chr.9: 130924465-130924465 on Build GRCh38
rs201227004 Chr.9: 130924414-130924414 on Build GRCh38
rs957570949 Chr.9: 130924378-130924378 on Build GRCh38
rs377485598 Chr.9: 130924351-130924351 on Build GRCh38
rs754297116 Chr.9: 130924401-130924401 on Build GRCh38
rs762141176 Chr.9: 130924367-130924367 on Build GRCh38
rs370732665 Chr.9: 130924360-130924360 on Build GRCh38
rs549728418 Chr.9: 130924438-130924438 on Build GRCh38
rs374186246 Chr.9: 130924400-130924400 on Build GRCh38
rs561385387 Chr.9: 130924420-130924420 on Build GRCh38
rs1017812284 Chr.9: 130924412-130924412 on Build GRCh38
rs565474779 Chr.9: 130924390-130924390 on Build GRCh38
rs563197613 Chr.9: 130924347-130924347 on Build GRCh38
rs748442332 Chr.9: 130924409-130924409 on Build GRCh38
rs999308157 Chr.9: 130924365-130924365 on Build GRCh38
rs34060074 Chr.9: 130924455-130924456 on Build GRCh38
rs1046726514 Chr.9: 130924520-130924520 on Build GRCh38
rs750548107 Chr.9: 130924385-130924385 on Build GRCh38
rs576776257 Chr.9: 130924352-130924352 on Build GRCh38
rs916498989 Chr.9: 130924466-130924466 on Build GRCh38

View Full Product Details