Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 134788619-134788892 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM122B
Entrez Gene ID
159090
Gene Name
family with sequence similarity 122B
Gene Aliases
SPACIA2
UniGene
Hs.404706
Gene Chromosome Location
Chr.X: 134769566-134797232 on Build GRCh38
Transcripts
NM_001166599.2 NM_001166600.2 NM_001170756.1 NM_001170757.1 NM_145284.5 XM_006724730.2 XM_006724732.2 XM_006724733.3 XM_011531282.1 XM_011531283.1 XM_011531284.1 XM_011531285.1 XM_011531286.1 XM_011531287.1 XM_011531288.1 XM_011531289.1 XM_011531290.1 XM_011531291.1 XM_011531292.1 XM_011531293.1 XM_011531294.1 XM_017029306.1 XM_017029307.1

SNP Detail

SNP ID Chromosome Location
rs761402998 Chr.X: 134788735-134788735 on Build GRCh38
rs772713945 Chr.X: 134788739-134788739 on Build GRCh38
rs771793362 Chr.X: 134788736-134788736 on Build GRCh38
rs368137082 Chr.X: 134788769-134788769 on Build GRCh38
rs201263467 Chr.X: 134788833-134788833 on Build GRCh38
rs879036889 Chr.X: 134788834-134788834 on Build GRCh38
rs758668432 Chr.X: 134788811-134788811 on Build GRCh38
rs768451437 Chr.X: 134788722-134788722 on Build GRCh38
rs746507504 Chr.X: 134788709-134788709 on Build GRCh38
rs747141847 Chr.X: 134788704-134788704 on Build GRCh38
rs749154850 Chr.X: 134788714-134788714 on Build GRCh38
rs371668938 Chr.X: 134788855-134788855 on Build GRCh38
rs375303160 Chr.X: 134788727-134788727 on Build GRCh38
rs765556053 Chr.X: 134788808-134788808 on Build GRCh38
rs145927426 Chr.X: 134788775-134788775 on Build GRCh38
rs770544570 Chr.X: 134788867-134788867 on Build GRCh38
rs749244627 Chr.X: 134788868-134788868 on Build GRCh38
rs777830558 Chr.X: 134788695-134788695 on Build GRCh38
rs371512792 Chr.X: 134788712-134788712 on Build GRCh38
rs764436411 Chr.X: 134788828-134788828 on Build GRCh38
rs760074336 Chr.X: 134788801-134788801 on Build GRCh38
rs754412406 Chr.X: 134788789-134788789 on Build GRCh38
rs753150171 Chr.X: 134788809-134788809 on Build GRCh38
rs770985244 Chr.X: 134788705-134788705 on Build GRCh38
rs368920471 Chr.X: 134788682-134788682 on Build GRCh38

View Full Product Details