Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 102731119-102731346 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC168
Entrez Gene ID
643677
Gene Name
coiled-coil domain containing 168
Gene Aliases
C13orf40
UniGene
Hs.368382
Gene Chromosome Location
Chr.13: 102729367-102759072 on Build GRCh38
Transcripts
NM_001146197.1 XM_011521106.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5349875 c.5498G>C p.R1833T Chr.13: 102731312-102731312 on Build GRCh38
6521208 c.5486G>T p.R1829M Chr.13: 102731324-102731324 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs958088988 Chr.13: 102731218-102731218 on Build GRCh38
rs970797011 Chr.13: 102731297-102731297 on Build GRCh38
rs749808704 Chr.13: 102731214-102731214 on Build GRCh38
rs990028031 Chr.13: 102731183-102731183 on Build GRCh38
rs1033330715 Chr.13: 102731208-102731208 on Build GRCh38
rs1039361401 Chr.13: 102731170-102731170 on Build GRCh38
rs558343130 Chr.13: 102731240-102731240 on Build GRCh38
rs781615454 Chr.13: 102731252-102731252 on Build GRCh38
rs752193306 Chr.13: 102731222-102731222 on Build GRCh38
rs977845385 Chr.13: 102731235-102731235 on Build GRCh38
rs750927809 Chr.13: 102731261-102731261 on Build GRCh38
rs928678720 Chr.13: 102731236-102731236 on Build GRCh38
rs759214848 Chr.13: 102731196-102731196 on Build GRCh38
rs773171442 Chr.13: 102731145-102731145 on Build GRCh38
rs757890156 Chr.13: 102731225-102731225 on Build GRCh38
rs536677293 Chr.13: 102731199-102731199 on Build GRCh38
rs1055920365 Chr.13: 102731256-102731256 on Build GRCh38
rs548119979 Chr.13: 102731158-102731158 on Build GRCh38
rs200341155 Chr.13: 102731207-102731207 on Build GRCh38
rs143423314 Chr.13: 102731262-102731262 on Build GRCh38
rs58979683 Chr.13: 102731147-102731147 on Build GRCh38
rs9514051 Chr.13: 102731174-102731174 on Build GRCh38

View Full Product Details