Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 56126881-56127150 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD55
Entrez Gene ID
79722
Gene Name
ankyrin repeat domain 55
Gene Aliases
-
UniGene
Hs.436214
Gene Chromosome Location
Chr.5: 56099680-56233359 on Build GRCh38
Transcripts
NM_024669.2 XM_011543646.1 XM_017009852.1 XM_017009853.1 XM_017009854.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7222240 c.687G>C p.E229D Chr.5: 56127032-56127032 on Build GRCh38
364662 c.643C>A p.L215M Chr.5: 56127076-56127076 on Build GRCh38
6442794 c.659G>A p.G220E Chr.5: 56127060-56127060 on Build GRCh38
3344727 c.717G>A p.A239A Chr.5: 56127002-56127002 on Build GRCh38
738486 c.776A>G p.Q259R Chr.5: 56126943-56126943 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753495263 Chr.5: 56127058-56127059 on Build GRCh38
rs780270426 Chr.5: 56126939-56126939 on Build GRCh38
rs748241548 Chr.5: 56126967-56126967 on Build GRCh38
rs35380800 Chr.5: 56126957-56126958 on Build GRCh38
rs201655144 Chr.5: 56127020-56127020 on Build GRCh38
rs372499783 Chr.5: 56127052-56127052 on Build GRCh38
rs946003276 Chr.5: 56126965-56126965 on Build GRCh38
rs61741177 Chr.5: 56126997-56126997 on Build GRCh38
rs940955090 Chr.5: 56127092-56127092 on Build GRCh38
rs375554059 Chr.5: 56126987-56126987 on Build GRCh38
rs763870042 Chr.5: 56127080-56127080 on Build GRCh38
rs891588692 Chr.5: 56126955-56126955 on Build GRCh38
rs781136419 Chr.5: 56126923-56126923 on Build GRCh38
rs201555961 Chr.5: 56126986-56126986 on Build GRCh38
rs61741176 Chr.5: 56126996-56126996 on Build GRCh38
rs146618027 Chr.5: 56127091-56127091 on Build GRCh38
rs777120326 Chr.5: 56126994-56126994 on Build GRCh38
rs749573028 Chr.5: 56126948-56126948 on Build GRCh38
rs751137611 Chr.5: 56127000-56127000 on Build GRCh38
rs771561192 Chr.5: 56127066-56127066 on Build GRCh38
rs763753117 Chr.5: 56126915-56126915 on Build GRCh38
rs755872832 Chr.5: 56126935-56126935 on Build GRCh38
rs773608846 Chr.5: 56127060-56127060 on Build GRCh38
rs751109287 Chr.5: 56126918-56126918 on Build GRCh38
rs2289857 Chr.5: 56127014-56127014 on Build GRCh38
rs747611870 Chr.5: 56127065-56127065 on Build GRCh38
rs760643375 Chr.5: 56127098-56127098 on Build GRCh38
rs757182743 Chr.5: 56126921-56126921 on Build GRCh38
rs772126737 Chr.5: 56127057-56127057 on Build GRCh38
rs200614650 Chr.5: 56127016-56127016 on Build GRCh38
rs752715295 Chr.5: 56127087-56127087 on Build GRCh38
rs750318320 Chr.5: 56127008-56127008 on Build GRCh38
rs749055439 Chr.5: 56127035-56127035 on Build GRCh38
rs773985846 Chr.5: 56127086-56127086 on Build GRCh38
rs748211795 Chr.5: 56127056-56127056 on Build GRCh38
rs745682018 Chr.5: 56126934-56126934 on Build GRCh38
rs766277805 Chr.5: 56127106-56127106 on Build GRCh38
rs763930496 Chr.5: 56126976-56126988 on Build GRCh38
rs777177240 Chr.5: 56127074-56127074 on Build GRCh38
rs113351727 Chr.5: 56127050-56127050 on Build GRCh38
rs1021223951 Chr.5: 56126990-56126990 on Build GRCh38
rs768864978 Chr.5: 56126953-56126953 on Build GRCh38
rs1054291767 Chr.5: 56126913-56126913 on Build GRCh38
rs541461564 Chr.5: 56127110-56127110 on Build GRCh38
rs1025828285 Chr.5: 56127070-56127070 on Build GRCh38
rs61742899 Chr.5: 56127096-56127096 on Build GRCh38
rs199752327 Chr.5: 56127003-56127003 on Build GRCh38
rs371960976 Chr.5: 56126960-56126960 on Build GRCh38
rs754732132 Chr.5: 56127121-56127121 on Build GRCh38
rs575822491 Chr.5: 56126979-56126979 on Build GRCh38
rs376402466 Chr.5: 56127002-56127002 on Build GRCh38

View Full Product Details