Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 88574784-88575053 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF2AK3
Entrez Gene ID
9451
Gene Name
eukaryotic translation initiation factor 2 alpha kinase 3
Gene Aliases
PEK, PERK, WRS
UniGene
Hs.591589
Gene Chromosome Location
Chr.2: 88556740-88627576 on Build GRCh38
Transcripts
NM_001313915.1 NM_004836.6 XM_017005376.1 NR_110236.1

Gene Symbol
LOC101928371
Entrez Gene ID
101928371
Gene Name
uncharacterized LOC101928371
Gene Aliases
-
UniGene
Hs.738045
Gene Chromosome Location
Chr.2: 88538720-88575610 on Build GRCh38
Transcripts
NM_001313915.1 NM_004836.6 XM_017005376.1 NR_110236.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4769954 c.2679A>G p.K893K Chr.2: 88574804-88574804 on Build GRCh38
3702253 c.2581A>C p.T861P Chr.2: 88574902-88574902 on Build GRCh38
5059109 c.2467A>G p.K823E Chr.2: 88575016-88575016 on Build GRCh38
5595779 c.2545_2547delTCT p.S849delS Chr.2: 88574936-88574938 on Build GRCh38
6010218 c.2570C>T p.P857L Chr.2: 88574913-88574913 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773451477 Chr.2: 88574931-88574931 on Build GRCh38
rs748587261 Chr.2: 88575017-88575017 on Build GRCh38
rs777452338 Chr.2: 88574810-88574810 on Build GRCh38
rs752633958 Chr.2: 88574972-88574972 on Build GRCh38
rs374540128 Chr.2: 88575016-88575016 on Build GRCh38
rs776415401 Chr.2: 88574959-88574959 on Build GRCh38
rs369602164 Chr.2: 88574949-88574949 on Build GRCh38
rs200955126 Chr.2: 88574898-88574898 on Build GRCh38
rs764889727 Chr.2: 88574969-88574969 on Build GRCh38
rs768762794 Chr.2: 88574913-88574913 on Build GRCh38
rs758593623 Chr.2: 88574936-88574938 on Build GRCh38
rs763883904 Chr.2: 88574979-88574979 on Build GRCh38
rs551274166 Chr.2: 88575018-88575018 on Build GRCh38
rs369837691 Chr.2: 88575000-88575000 on Build GRCh38
rs780380140 Chr.2: 88574897-88574897 on Build GRCh38
rs137868793 Chr.2: 88574974-88574974 on Build GRCh38
rs56094918 Chr.2: 88574951-88574951 on Build GRCh38
rs746064190 Chr.2: 88574850-88574850 on Build GRCh38
rs765936619 Chr.2: 88574952-88574952 on Build GRCh38
rs751296708 Chr.2: 88574983-88574983 on Build GRCh38
rs150135027 Chr.2: 88574928-88574928 on Build GRCh38
rs769915392 Chr.2: 88574870-88574870 on Build GRCh38
rs375265921 Chr.2: 88574927-88574927 on Build GRCh38
rs757139632 Chr.2: 88574987-88574987 on Build GRCh38
rs767541817 Chr.2: 88575021-88575023 on Build GRCh38
rs755001234 Chr.2: 88575007-88575007 on Build GRCh38
rs759089652 Chr.2: 88574962-88574962 on Build GRCh38
rs772383156 Chr.2: 88574930-88574930 on Build GRCh38
rs200270016 Chr.2: 88574976-88574976 on Build GRCh38

View Full Product Details