Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 43407361-43407616 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HECW1
Entrez Gene ID
23072
Gene Name
HECT, C2 and WW domain containing E3 ubiquitin protein ligase 1
Gene Aliases
NEDL1
UniGene
Hs.164453
Gene Chromosome Location
Chr.7: 43112580-43566001 on Build GRCh38
Transcripts
NM_001287059.1 NM_015052.4 XM_005249665.3 XM_006715670.3 XM_006715671.3 XM_006715673.3 XM_011515220.2 XM_011515222.2 XM_011515223.2 XM_011515224.2 XM_011515225.2 XM_011515226.2 XM_017011882.1 XM_017011883.1 XM_017011884.1 XM_017011885.1 XM_017011886.1 XM_017011887.1 XM_017011888.1 XM_017011889.1 XM_017011890.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1254070 c.653A>C p.K218T Chr.7: 43407583-43407583 on Build GRCh38
4455108 c.671A>G p.N224S Chr.7: 43407601-43407601 on Build GRCh38
4822124 c.658G>C p.G220R Chr.7: 43407588-43407588 on Build GRCh38
1450777 c.664T>C p.F222L Chr.7: 43407594-43407594 on Build GRCh38
3638746 c.680C>T p.P227L Chr.7: 43407610-43407610 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777991239 Chr.7: 43407573-43407573 on Build GRCh38
rs925315766 Chr.7: 43407422-43407422 on Build GRCh38
rs754368378 Chr.7: 43407544-43407544 on Build GRCh38
rs1037173373 Chr.7: 43407399-43407399 on Build GRCh38
rs895951880 Chr.7: 43407460-43407460 on Build GRCh38
rs950138210 Chr.7: 43407474-43407474 on Build GRCh38
rs949263402 Chr.7: 43407503-43407503 on Build GRCh38
rs766802292 Chr.7: 43407531-43407531 on Build GRCh38
rs558575036 Chr.7: 43407391-43407391 on Build GRCh38
rs758712422 Chr.7: 43407562-43407562 on Build GRCh38
rs938088860 Chr.7: 43407452-43407452 on Build GRCh38
rs765678441 Chr.7: 43407554-43407554 on Build GRCh38
rs866036926 Chr.7: 43407567-43407567 on Build GRCh38
rs910159230 Chr.7: 43407390-43407390 on Build GRCh38
rs780362783 Chr.7: 43407484-43407484 on Build GRCh38
rs577021472 Chr.7: 43407513-43407513 on Build GRCh38
rs979431165 Chr.7: 43407419-43407419 on Build GRCh38
rs551039763 Chr.7: 43407475-43407475 on Build GRCh38
rs753067295 Chr.7: 43407558-43407558 on Build GRCh38
rs550921490 Chr.7: 43407512-43407512 on Build GRCh38
rs201174011 Chr.7: 43407584-43407584 on Build GRCh38
rs866714710 Chr.7: 43407581-43407581 on Build GRCh38
rs755455675 Chr.7: 43407546-43407546 on Build GRCh38
rs966666420 Chr.7: 43407409-43407409 on Build GRCh38

View Full Product Details