Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 107670462-107670709 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFNA5
Entrez Gene ID
1946
Gene Name
ephrin A5
Gene Aliases
AF1, EFL5, EPLG7, GLC1M, LERK7, RAGS
UniGene
Hs.288741
Gene Chromosome Location
Chr.5: 107376889-107672364 on Build GRCh38
Transcripts
NM_001962.2 XM_006714565.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1753898 c.60C>T p.S20S Chr.5: 107670554-107670554 on Build GRCh38
1059355 c.110A>G p.N37S Chr.5: 107670504-107670504 on Build GRCh38
4656577 c.95A>T p.Y32F Chr.5: 107670519-107670519 on Build GRCh38
1753899 c.57C>T p.F19F Chr.5: 107670557-107670557 on Build GRCh38
1059356 c.12G>A p.V4V Chr.5: 107670602-107670602 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375348285 Chr.5: 107670573-107670573 on Build GRCh38
rs754892007 Chr.5: 107670574-107670574 on Build GRCh38
rs147882024 Chr.5: 107670491-107670491 on Build GRCh38
rs765473063 Chr.5: 107670515-107670515 on Build GRCh38
rs377351448 Chr.5: 107670667-107670667 on Build GRCh38
rs767936434 Chr.5: 107670636-107670636 on Build GRCh38
rs976533069 Chr.5: 107670681-107670682 on Build GRCh38
rs897987874 Chr.5: 107670554-107670554 on Build GRCh38
rs553936811 Chr.5: 107670483-107670483 on Build GRCh38
rs202114413 Chr.5: 107670561-107670561 on Build GRCh38
rs751892462 Chr.5: 107670536-107670536 on Build GRCh38
rs980162174 Chr.5: 107670686-107670686 on Build GRCh38
rs750133350 Chr.5: 107670569-107670569 on Build GRCh38
rs775763363 Chr.5: 107670628-107670628 on Build GRCh38
rs778726581 Chr.5: 107670656-107670656 on Build GRCh38
rs764499773 Chr.5: 107670533-107670533 on Build GRCh38
rs368040477 Chr.5: 107670624-107670624 on Build GRCh38
rs774711441 Chr.5: 107670632-107670632 on Build GRCh38
rs947444345 Chr.5: 107670672-107670672 on Build GRCh38
rs776985020 Chr.5: 107670620-107670620 on Build GRCh38
rs370590979 Chr.5: 107670526-107670526 on Build GRCh38
rs748177986 Chr.5: 107670657-107670657 on Build GRCh38
rs755615518 Chr.5: 107670571-107670571 on Build GRCh38
rs764319874 Chr.5: 107670656-107670657 on Build GRCh38
rs570824137 Chr.5: 107670644-107670644 on Build GRCh38
rs548463016 Chr.5: 107670630-107670630 on Build GRCh38
rs753373791 Chr.5: 107670652-107670652 on Build GRCh38
rs371484447 Chr.5: 107670631-107670631 on Build GRCh38
rs1028630487 Chr.5: 107670614-107670614 on Build GRCh38
rs771131134 Chr.5: 107670609-107670609 on Build GRCh38
rs769965845 Chr.5: 107670625-107670625 on Build GRCh38
rs531625106 Chr.5: 107670606-107670606 on Build GRCh38
rs201171573 Chr.5: 107670637-107670637 on Build GRCh38
rs199980747 Chr.5: 107670512-107670512 on Build GRCh38
rs762070982 Chr.5: 107670635-107670635 on Build GRCh38
rs866795451 Chr.5: 107670497-107670497 on Build GRCh38
rs756893997 Chr.5: 107670547-107670547 on Build GRCh38
rs200187971 Chr.5: 107670640-107670640 on Build GRCh38
rs756694630 Chr.5: 107670590-107670590 on Build GRCh38
rs754874895 Chr.5: 107670653-107670653 on Build GRCh38
rs898892115 Chr.5: 107670556-107670556 on Build GRCh38
rs763595655 Chr.5: 107670527-107670527 on Build GRCh38
rs758128708 Chr.5: 107670664-107670664 on Build GRCh38
rs954364490 Chr.5: 107670619-107670619 on Build GRCh38

View Full Product Details