Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 37361963-37362236 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HKR1
Entrez Gene ID
284459
Gene Name
HKR1, GLI-Kruppel zinc finger family member
Gene Aliases
ZNF875
UniGene
Hs.657027
Gene Chromosome Location
Chr.19: 37317900-37364455 on Build GRCh38
Transcripts
NM_181786.2 XM_017026672.1 XM_017026673.1 XM_017026674.1 XM_017026675.1 XM_017026676.1 XM_017026677.1 XM_017026678.1 XM_017026679.1 XM_017026680.1 XM_017026681.1 XM_017026682.1 XM_017026683.1 XM_017026684.1 XM_017026685.1 XM_017026686.1 XM_017026687.1 XM_017026688.1 XM_017026689.1 XM_017026690.1 XM_017026691.1 XM_017026692.1 XM_017026693.1 XM_017026694.1 XM_017026695.1 XM_017026696.1 XM_017026697.1 XM_017026698.1 XM_017026699.1 XM_017026700.1 XM_017026701.1 XM_017026702.1 XM_017026703.1 XM_017026704.1 XM_017026705.1 XM_017026707.1 XM_017026708.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4391701 c.419C>A p.S140* Chr.19: 37362214-37362214 on Build GRCh38
3822873 c.268G>C p.E90Q Chr.19: 37362120-37362120 on Build GRCh38
5921435 c.284C>T p.P95L Chr.19: 37362136-37362136 on Build GRCh38
4490456 c.306C>T p.S102S Chr.19: 37362158-37362158 on Build GRCh38
439355 c.314-1G>A p.? Chr.19: 37362108-37362108 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781366203 Chr.19: 37362163-37362163 on Build GRCh38
rs534877758 Chr.19: 37362032-37362032 on Build GRCh38
rs757153287 Chr.19: 37362080-37362080 on Build GRCh38
rs762991817 Chr.19: 37362090-37362090 on Build GRCh38
rs922386960 Chr.19: 37361997-37361997 on Build GRCh38
rs148162251 Chr.19: 37362113-37362113 on Build GRCh38
rs200796429 Chr.19: 37362184-37362184 on Build GRCh38
rs932376484 Chr.19: 37362041-37362041 on Build GRCh38
rs775596561 Chr.19: 37362083-37362083 on Build GRCh38
rs201374559 Chr.19: 37362157-37362157 on Build GRCh38
rs746224331 Chr.19: 37362179-37362179 on Build GRCh38
rs781597721 Chr.19: 37362128-37362128 on Build GRCh38
rs751673317 Chr.19: 37362099-37362099 on Build GRCh38
rs199540276 Chr.19: 37362202-37362202 on Build GRCh38
rs1057010686 Chr.19: 37362209-37362209 on Build GRCh38
rs771146877 Chr.19: 37362057-37362057 on Build GRCh38
rs376490409 Chr.19: 37362145-37362145 on Build GRCh38
rs768459309 Chr.19: 37362015-37362018 on Build GRCh38
rs545227345 Chr.19: 37362026-37362026 on Build GRCh38
rs746754947 Chr.19: 37362076-37362077 on Build GRCh38
rs372946836 Chr.19: 37362103-37362103 on Build GRCh38
rs370252285 Chr.19: 37362063-37362063 on Build GRCh38
rs965574894 Chr.19: 37362005-37362005 on Build GRCh38
rs977361268 Chr.19: 37362006-37362006 on Build GRCh38
rs774660410 Chr.19: 37362206-37362206 on Build GRCh38
rs777204267 Chr.19: 37362069-37362069 on Build GRCh38
rs1056838819 Chr.19: 37362185-37362185 on Build GRCh38
rs747466985 Chr.19: 37362056-37362056 on Build GRCh38
rs554577004 Chr.19: 37362171-37362171 on Build GRCh38
rs564254965 Chr.19: 37362072-37362072 on Build GRCh38
rs769885438 Chr.19: 37362076-37362076 on Build GRCh38
rs370976349 Chr.19: 37362148-37362148 on Build GRCh38
rs764355928 Chr.19: 37362097-37362097 on Build GRCh38
rs754524650 Chr.19: 37362152-37362152 on Build GRCh38
rs939375199 Chr.19: 37362174-37362174 on Build GRCh38
rs763031022 Chr.19: 37362196-37362196 on Build GRCh38
rs561506730 Chr.19: 37362158-37362158 on Build GRCh38
rs111387020 Chr.19: 37362211-37362211 on Build GRCh38
rs373644320 Chr.19: 37362112-37362112 on Build GRCh38

View Full Product Details