Gene Transcripts

Gene Symbol
Pwp2
Entrez Gene ID
110816
Gene Name
PWP2 periodic tryptophan protein homolog (yeast)
Gene Aliases
6530411D08Rik, Pwp2h, wdp103
Chromosome Location
Chr. 10: 78170910 - 78185171 on Build GRCm38
UniGene ID
Mm.103522
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029546.2 NP_083822.1 13 1575
GenBank mRNA AB041855.1 - 13 1536
AK088254.1 - 13 1576
AK088983.1 - 13 1576
BC031787.1 - 2 62
BC112325.1 - 13 1541 5355810
BC128480.1 - 13 1430 40097069

View Full Product Details