Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_054066.4 | NP_473407.2 | 8 | 1235 | |
GenBank mRNA | AF435950.1 | - | 8 | 1240 | |
AK006672.1 | - | 8 | 1280 | ||
BC106767.1 | - | 8 | 1230 | 40039566 | |
BC106768.1 | - | 8 | 1233 | 40039567 |