Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_009626.4 | NP_033756.2 | 6, 7 | 953 | |
GenBank mRNA | AF331802.1 | - | 6, 7 | 951 | |
AF331803.1 | - | 6, 7 | 951 | ||
AK009588.2 | - | 6, 7 | 920 | ||
AK009600.1 | - | 6, 7 | 966 | ||
AK144795.1 | - | 904 | |||
BC047267.1 | - | 6, 7 | 882 | 6310424 | |
U20257.1 | - | 6, 7 | 844 |