Gene Transcripts

Gene Symbol
Adsl
Entrez Gene ID
11564
Gene Name
adenylosuccinate lyase
Gene Aliases
Adl, Asl
Chromosome Location
Chr. 15: 80948490 - 80970947 on Build GRCm38
UniGene ID
Mm.38151
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009634.6 NP_033764.2 2 253
GenBank mRNA AK049372.1 - 2 225
AK051111.1 - 2 225
AK152092.1 - 2 225
AK166019.1 - 2 221
AK168906.1 - 2 218
AK170216.1 - 2 218
AK171725.1 - 2 234
BC020187.1 - 2 229 4913637
U20225.1 - 2 229

View Full Product Details