Gene Transcripts

Gene Symbol
Ambp
Entrez Gene ID
11699
Gene Name
alpha 1 microglobulin/bikunin
Gene Aliases
AI194774, ASPI, HI-30, Intin4, Itil, UTI
Chromosome Location
Chr. 4: 63143279 - 63154142 on Build GRCm38
UniGene ID
Mm.2197
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007443.3 NP_031469.1 9 1013
GenBank mRNA AK004907.1 - 9 1014
BC021660.1 - 9 1032 4193922
D28812.1 - 9 1014
X68680.1 - 9 975

View Full Product Details